Merck
  • Protein labeling with the labeling precursor [(18)F]SiFA-SH for positron emission tomography.
Protein labeling with the labeling precursor [(18)F]SiFA-SH for positron emission tomography.
Nature protocols (2012-10-06)
Björn Wängler, Alexey P Kostikov, Sabrina Niedermoser, Joshua Chin, Katy Orchowski, Esther Schirrmacher, Liuba Iovkova-Berends, Klaus Jurkschat, Carmen Wängler, Ralf Schirrmacher
ABSTRACT
Proteins previously derivatized with the cross-coupling reagent sulfo-SMCC (4-(N-maleimidomethyl)cyclohexane-1-carboxylic acid 3-sulfo-N-hydroxy-succinimide ester sodium salt) can be easily labeled in high radiochemical yields with the silicon-fluoride acceptor (SiFA) reagent [(18)F]SiFA-SH, obtained via isotopic exchange, by thiol-maleimide coupling chemistry (n = 10). The specific activity of SiFA-SH obtained in a one-step labeling reaction was > 18.5 GBq μmol(-1) (> 500 Ci mmol(-1)). The number of SiFA building blocks per protein molecule is defined by the previously introduced number of maleimide groups, which can be determined by a simple and convenient Ellman's assay. Not more than two maleimide groups are introduced using sulfo-SMCC, thereby keeping the modification of the protein low and preserving its biological activity.
Sigma-Aldrich
Albumin from rat serum, lyophilized powder, ≥96% (agarose gel electrophoresis)
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%