Merck
  • The cell death protease Kex1p is essential for hypochlorite-induced apoptosis in yeast.
The cell death protease Kex1p is essential for hypochlorite-induced apoptosis in yeast.
Cell cycle (Georgetown, Tex.) (2013-05-10)
Didac Carmona-Gutierrez, Ali Alavian-Ghavanini, Lukas Habernig, Maria Anna Bauer, Astrid Hammer, Christine Rossmann, Andreas S Zimmermann, Christoph Ruckenstuhl, Sabrina Büttner, Tobias Eisenberg, Wolfgang Sattler, Ernst Malle, Frank Madeo
ABSTRACT
Following microbial pathogen invasion, the human immune system of activated phagocytes generates and releases the potent oxidant hypochlorous acid (HOCl), which contributes to the killing of menacing microorganisms. Though tightly controlled, HOCl generation by the myeloperoxidase-hydrogen peroxide-chloride system of neutrophils/monocytes may occur in excess and lead to tissue damage. It is thus of marked importance to delineate the molecular pathways underlying HOCl cytotoxicity in both microbial and human cells. Here, we show that HOCl induces the generation of reactive oxygen species (ROS), apoptotic cell death and the formation of specific HOCl-modified epitopes in the budding yeast Saccharomyces cerevisiae. Interestingly, HOCl cytotoxicity can be prevented by treatment with ROS scavengers, suggesting oxidative stress to mediate the lethal effect. The executing pathway involves the pro-apoptotic protease Kex1p, since its absence diminishes HOCl-induced production of ROS, apoptosis and protein modification. By characterizing HOCl-induced cell death in yeast and identifying a corresponding central executor, these results pave the way for the use of Saccharomyces cerevisiae in HOCl research, not least given that it combines both being a microorganism as well as a model for programmed cell death in higher eukaryotes.
Sigma-Aldrich
Dihydroethidium, ≥95%
Sigma-Aldrich
N-Acetyl-D-sphingosine, ≥97% (TLC), powder
Roche
In Situ Cell Death Detection Kit, Fluorescein, sufficient for ≤50 tests, suitable for detection