Merck
  • High-resolution analysis of genetic stability of human adipose tissue stem cells cultured to senescence.
High-resolution analysis of genetic stability of human adipose tissue stem cells cultured to senescence.
Journal of cellular and molecular medicine (2008-04-19)
Leonardo A Meza-Zepeda, Agate Noer, John Arne Dahl, Francesca Micci, Ola Myklebost, Philippe Collas
ABSTRACT
The potential use of human mesenchymal stem cells for therapeutic applications implies large scale in vitro culture, increasing the probability of genetic instability and transformation. We examine here the incidence of unbalanced and balanced chromosome rearrangements in polyclonal and single cell-derived cultures of human adipose stem cells to senescence. G-banding karyotyping of the polyclonal cultures shows a normal karyotype. In addition, high-resolution microarray-based comparative genomic hybridization analyses relative to uncultured adipose stem cells from the same donors reveal overall genomic stability in long-term (approximately 6 months) polyclonal and clonal culture. One adipose stem cell clone displayed minor deletions in gene-rich telomeric and sub-telomeric regions on three chromosomes in early passage. This however, was detected only in a sub-population of cells that was subsequently spontaneously eliminated from the culture. Apparent pericentromeric instabilities are also occasionally detected in specific chromosomes. Our results indicate that clonal chromosomal aberrations may arise transiently in early passage adipose stem cells (ASC) cultures. Nonetheless, incidence of these aberrations seems to be negligible in the majority of long-term ASC cultures, at least under the culture conditions used here.
Sigma-Aldrich
NAD Kinase human, recombinant, expressed in E. coli, ≥95% (SDS-PAGE)