Merck
  • Expression, purification, crystallization and preliminary X-ray analysis of CttA, a putative cellulose-binding protein from Ruminococcus flavefaciens.
Expression, purification, crystallization and preliminary X-ray analysis of CttA, a putative cellulose-binding protein from Ruminococcus flavefaciens.
Acta crystallographica. Section F, Structural biology communications (2015-06-10)
Immacolata Venditto, Pedro Bule, Andrew Thompson, Juan Sanchez-Weatherby, James Sandy, Luis M A Ferreira, Carlos M G A Fontes, Shabir Najmudin
ABSTRACT
A number of anaerobic microorganisms produce multi-modular, multi-enzyme complexes termed cellulosomes. These extracellular macromolecular nanomachines are designed for the efficient degradation of plant cell-wall carbohydrates to smaller sugars that are subsequently used as a source of carbon and energy. Cellulolytic strains from the rumens of mammals, such as Ruminococcus flavefaciens, have been shown to have one of the most complex cellulosomal systems known. Cellulosome assembly requires the binding of dockerin modules located in cellulosomal enzymes to cohesin modules located in a macromolecular scaffolding protein. Over 220 genes encoding dockerin-containing proteins have been identified in the R. flavefaciens genome. The dockerin-containing enzymes can be incorporated into the primary scaffoldin (ScaA), which in turn can bind to adaptor scaffoldins (ScaB or ScaC) and subsequently to anchoring scaffoldin (ScaE), thereby attaching the whole complex to the cell surface. However, unlike other cellulosomes such as that from Clostridium thermocellum, the Ruminococcus species lack a specific carbohydrate-binding module (CBM) on ScaA which recruits the entire complex onto the surface of the substrate. Instead, a cellulose-binding protein, CttA, comprising two putative tandem novel carbohydrate-binding modules and a C-terminal X-dockerin module, which can bind to the cohesin of ScaE, may mediate the attachment of bacterial cells to cellulose. Here, the expression, purification and crystallization of the carbohydrate-binding modular part of the CttA from R. flavefaciens are described. X-ray data have been collected to resolutions of 3.23 and to 1.61 Å in space groups P3(1)21 or P3(2)21 and P2(1), respectively. The structure was phased using bound iodide from the crystallization buffer by SAD experiments.
Product Description
SAFC
Sodium chloride solution, 5 M
SAFC
HEPES
Sigma-Aldrich
Sodium chloride solution, 0.85%
SAFC
HEPES
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
HEPES, BioUltra, for molecular biology, ≥99.5% (T)
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.9% trace metals basis
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.99% trace metals basis
Sigma-Aldrich
Calcium chloride solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
Sodium iodide, ≥99.99% trace metals basis
Sigma-Aldrich
Sodium iodide, 99.999% trace metals basis