Merck
  • Antidepressant/anxiolytic potential and adverse effect liabilities of melanin-concentrating hormone receptor 1 antagonists in animal models.
Antidepressant/anxiolytic potential and adverse effect liabilities of melanin-concentrating hormone receptor 1 antagonists in animal models.
Pharmacology, biochemistry, and behavior (2015-06-06)
Shigeyuki Chaki, Toshiharu Shimazaki, Mariko Nishiguchi, Takeo Funakoshi, Michihiko Iijima, Akie Ito, Kosuke Kanuma, Yoshinori Sekiguchi
ABSTRACT
Melanin-concentrating hormone receptor 1 (MCH1 receptor) is known to be involved in the control of mood and stress, in addition to the regulation of feeding. Here, we report further evidence that the blockade of the MCH1 receptor exhibits antidepressant and anxiolytic-like effects in a variety of animal models using TASP0382650 and TASP0489838, newly synthesized MCH1 receptor antagonists, with different scaffolds. Both TASP0382650 and TASP0489838 exhibited high affinities for human MCH1 receptor with IC50 values of 7.13 and 3.80nM, respectively. Both compounds showed potent antagonist activities at the MCH1 receptor, as assessed using MCH-increased [(35)S]GTPγS binding to human MCH1 receptor and an MCH-induced [Ca(2+)]i assay in rat MCH1 receptor expressing cells. In contrast, neither TASP0382650 nor TASP0489838 showed an affinity for the MCH2 receptor, another MCH receptor subtype. The oral administration of TASP0382650 or TASP0489838 significantly reduced the immobility time during the forced swimming test in rats, and reduced hyperemotionality induced by an olfactory bulbectomy, both of which are indicative of an antidepressant-like potential. In the olfactory bulbectomy model, the antidepressant effect of TASP0382650 appeared following a single administration, suggesting a faster onset of action, compared with current medications. Moreover, both TASP0382650 and TASP0489838 exhibited anxiolytic effects in several animal models of anxiety. In contrast, both TASP0382650 and TASP0489838 did not affect spontaneous locomotor activity, motor function, spatial memory during the Morris water maze task, or the convulsion threshold to pentylenetetrazole. These findings provide additional evidence that the blockade of the MCH1 receptor exhibits antidepressant- and anxiolytic activities with no adverse effects in experimental animal models.
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ≥99.0% (RT)
Sigma-Aldrich
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
L-Ascorbic acid, FCC, FG
Sigma-Aldrich
1-Hexanethiol, 95%
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
L-Ascorbic acid, meets USP testing specifications
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Sigma-Aldrich
L-Ascorbic acid, reagent grade
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Sigma-Aldrich
L-Ascorbic acid, 99%
Sigma-Aldrich
L-Ascorbic acid, BioUltra, ≥99.5% (RT)
Sigma-Aldrich
1-Hexanethiol, 99% (GC)
Sigma-Aldrich
Calcium chloride
Sigma-Aldrich
Calcium chloride, anhydrous, BioReagent, suitable for insect cell culture, suitable for plant cell culture, ≥96.0%
Sigma-Aldrich
Calcium chloride solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.9% trace metals basis
Sigma-Aldrich
Calcium chloride, anhydrous, powder, 99.99% trace metals basis
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.99% trace metals basis
Sigma-Aldrich
Calcium chloride, Vetec, reagent grade, 96%