Merck
  • Unimolecular dissociation of anions derived from maleic acid (MaH2) in the gas phase: MaH- and MaMgCl--- relationship to Grignard chemistry and reductive CO2 fixation.
Unimolecular dissociation of anions derived from maleic acid (MaH2) in the gas phase: MaH- and MaMgCl--- relationship to Grignard chemistry and reductive CO2 fixation.
European journal of mass spectrometry (Chichester, England) (2015-08-27)
Glenn B S Miller, Vincent Fäseke, Einar Uggerud
ABSTRACT
We have conducted collision induced dissociation experiments on the hydrogen maleate anion (MaH(-), m/z = 115) and the anionic maleate MgCl complex (MaMgC(-), m/z = 173). In addition, we have computationally investigated the observed fragmentation reactions. We find that both anions readily undergo two consecutive decarboxylations resulting in product ions at m/z = 71 and 27 for MaH(-), and at m/z = 129 and 85 for MaMgCl(-). The first decarboxylation is more facile for MaMgCl(-) than for MaH(-), while loss of CO(2) from Ma(-CO(2))H(-) is more facile than for Ma(-CO(2))MgCl(-). We also find that MaH(-) loses water, and we propose a mechanism for this loss. No first-generation fragmentation product other than Ma(-CO(2))MgCl(-) is seen for MaMgCl(-). Based on the observed unimolecular chemistry, we discuss some of its implications on reductive CO(2)-fixation and Grignard chemistry.
Sigma-Aldrich
Magnesium chloride solution, 0.1 M
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, ~0.025 M in H2O
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, 2 M in H2O
Sigma-Aldrich
Maleic anhydride, 99%
Sigma-Aldrich
Magnesium chloride solution, PCR Reagent, 25 mM MgCI2 solution for PCR
Sigma-Aldrich
Magnesium chloride solution, for molecular biology, 1.00 M±0.01 M
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.9% trace metals basis
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Magnesium chloride, powder, <200 μm
Sigma-Aldrich
Magnesium chloride, anhydrous, ≥98%
Sigma-Aldrich
Magnesium chloride, BioReagent, suitable for insect cell culture, ≥97.0%