Merck
Origin assessment of EV olive oils by esterified sterols analysis.
Food chemistry (2015-06-05)
Rosa Giacalone, Salvatore Giuliano, Eleonora Gulotta, Maria Monfreda, Giovanni Presti
ABSTRACT
In this study extra virgin olive oils of Italian and non-Italian origin (from Spain, Tunisia and blends of EU origin) were differentiated by GC-FID analysis of sterols and esterified sterols followed by chemometric tools. PCA allowed to highlight the high significance of esterified sterols to characterise extra virgin olive oils in relation to their origin. SIMCA provided a sensitivity and specificity of 94.39% and 91.59% respectively; furthermore, an external set of 54 extra virgin olive oils bearing a designation of Italian origin on the labelling was tested by SIMCA. Prediction results were also compared with organoleptic assessment. Finally, the poor correlation found between ethylesters and esterified sterols allowed to hazard the guess, worthy of further investigations, that esterified sterols may prove to be promising in studies of geographical discrimination: indeed they appear to be independent of those factors causing the formation of ethyl esters and related to olive oil production.
Sigma-Aldrich
Palmitic acid, ≥98%, FCC, FG
Sigma-Aldrich
Palmitic acid, ≥98% palmitic acid basis (GC)
Sigma-Aldrich
Dodecyl arachidate, ≥99%
Sigma-Aldrich
Palmitic acid, ≥99%
Sigma-Aldrich
Palmitic acid, BioXtra, ≥99%
Sigma-Aldrich
Palmitic acid, natural, 98%, FG
Sigma-Aldrich
Oleic acid, natural, FCC
Sigma-Aldrich
Oleic acid, technical grade, 90%
Sigma-Aldrich
Oleic acid, BioReagent, suitable for cell culture
Sigma-Aldrich
Oleic acid, ≥99% (GC)
Sigma-Aldrich
Oleic acid, meets analytical specification of Ph, Eur., 65.0-88.0% (GC)