Merck
  • Stimulation of Proliferation and Migration of Mouse Macrophages by Type B CpG-ODNs Is F-Spondin and IL-1Ra Dependent.
Stimulation of Proliferation and Migration of Mouse Macrophages by Type B CpG-ODNs Is F-Spondin and IL-1Ra Dependent.
PloS one (2015-06-05)
Tai-An Chen, Chiao-Chun Liao, Yung-Chih Cheng, Yen-Po Chen, Yi-Fan Hsu, Chi-Ming Liang, Shu-Mei Liang
ABSTRACT
Macrophage proliferation and migration are important for many facets of immune response. Here we showed that stimulation of macrophages with type B CpG oligodeoxynucleotides (CpG-B ODNs) such as CpG-ODN 1668 increased the production of anti-inflammatory cytokine interleukin 1 receptor antagonist (IL-1Ra) in a TLR9- and MyD88-dependent manner. The CpG-B ODNs-induced IL-1Ra increased macrophage migration and promoted macrophage proliferation by down-regulating the expression of a cell cycle negative regulator, p27 to increase cell population in the S phase. The induction of IL-1Ra by CpG-B ODNs was F-spondin dependent. Knockdown of F-spondin and IL-1Ra decreased CpG-B ODNs-induced macrophage migration whereas overexpression of IL-1Ra increased migration of those cells. These findings demonstrated novel roles for F-spondin and IL-1Ra in CpG-B ODNs-mediated cell proliferation and migration of macrophages.
Sigma-Aldrich
L-Glutamine
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
HEPES buffer solution, 1 M in H2O
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
SAFC
L-Glutamine
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
Supelco
Ethanol standards 10% (v/v), 10 % (v/v) in H2O, analytical standard
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
HEPES, BioUltra, for molecular biology, ≥99.5% (T)
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Spon2
Sigma-Aldrich
MISSION® esiRNA, targeting human SPON1
SAFC
HEPES
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%
SAFC
HEPES