Merck
  • High-performance liquid chromatographic enantioseparation of amino alcohol analogues possessing 1,2,3,4-tetrahydroisoquinoline skeleton on polysaccharide-based chiral stationary phases.
High-performance liquid chromatographic enantioseparation of amino alcohol analogues possessing 1,2,3,4-tetrahydroisoquinoline skeleton on polysaccharide-based chiral stationary phases.
Biomedical chromatography : BMC (2014-10-24)
Nóra Grecsó, István Ilisz, Zsanett Gecse, László Schönstein, Ferenc Fülöp, Antal Péter
ABSTRACT
The stereoisomers of 1,2,3,4-tetrahydroisoquinoline amino alcohol analogues and derivatives thereof were separated in normal-phase mode on chiral stationary phases based on preprepared silica coated with cellulose tris-(3,5-dimethylphenyl carbamate), cellulose tris-(3-chloro-4-methylphenyl carbamate), cellulose tris-(4-methylbenzoate) or cellulose tris-(4-chloro-3-methylphenyl carbamate). On all the investigated chiral columns, the retention and the enantioseparation were influenced by the nature and the concentrations of the mobile phase components and additives, and also the temperature. Experiments were performed in the temperature range 10-50°C. Thermodynamic parameters were calculated from plots of lnα vs 1/T. On these polysaccharide-based chiral columns, both enthalpy-driven separations and entropy-controlled enantioseparations were observed. The latter was advantageous with regard to the shorter retention and greater selectivity at high temperature. The sequence of elution of the stereoisomers was determined in all cases.
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Heptane, anhydrous, 99%
Sigma-Aldrich
1-Propanol, anhydrous, 99.7%
Sigma-Aldrich
1-Propanol, ≥99%, FG
Sigma-Aldrich
Isopropyl alcohol, ≥99.7%, FCC, FG
Sigma-Aldrich
1-Propanol, natural, ≥98%, FG
Sigma-Aldrich
1-Butanol, for molecular biology, ≥99%
Sigma-Aldrich
1-Butanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Hexane, HPLC Plus, for HPLC, GC, and residue analysis, ≥95%
Supelco
Methanol solution, contains 0.10 % (v/v) formic acid, UHPLC, suitable for mass spectrometry (MS), ≥99.5%
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
2-Propanol, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
tert-Butanol, anhydrous, ≥99.5%
Sigma-Aldrich
2-Propanol, anhydrous, 99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Butyl alcohol, natural, ≥99.5%, FCC, FG
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Hexane, anhydrous, 95%
Sigma-Aldrich
2-Propanol, ACS reagent, ≥99.5%
Supelco
Ethanol standards 10% (v/v), 10 % (v/v) in H2O, analytical standard
Sigma-Aldrich
2-Propanol, for molecular biology, BioReagent, ≥99.5%
Sigma-Aldrich
2-Propanol, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%, poly coated bottles
Sigma-Aldrich
2-Propanol, BioUltra, for molecular biology, ≥99.5% (GC)