Merck
  • Synthesis and characterization of self-curing hydrophilic bone cements for protein delivery.
Synthesis and characterization of self-curing hydrophilic bone cements for protein delivery.
Journal of biomedical materials research. Part B, Applied biomaterials (2014-09-12)
E Franco-Marquès, J Parra, M A Pèlach, J A Méndez
ABSTRACT
New formulations of acrylic bone cements for bone defect reparation, based on self-hardening methyl methacrylate (MMA)/methacrylic acid (MAA), with a high capacity for protein delivery, have been developed. The self-curing formulations were prepared by partial substitution of solid phase PMMA microparticles by newly obtained PMAA microspheres. The PMAA microspheres were prepared by inverse suspension polymerization of their monomer and were cross-linked with N,N'-methylene-bis-acrylamide (MBA) (10-15 wt %) to produce stable systems in contact with aqueous media. PMAA microspheres were loaded with hydrolyzed collagen (HC) as a model protein to simulate bone morphogenetic protein delivery useful for hard tissue reconstruction. Solid phase PMMA microparticles in the formulation were partially substituted by new PMAA-HC microspheres and were characterized to determine viability as an acrylic bone cement in minimally invasive surgery. The incorporation of PMAA-HC microspheres decreased peak temperature by 20°C, which minimized thermal necrotic risk after implantation. Mechanical compression tests revealed a behavior, under dry conditions, close to ISO 5833 standard requirements. However, a drastic drop in mechanical strength, ∼64%, was obtained after 15 days of immersion in simulated physiological conditions (37°C and pH 7.4) and was attributed to water absorption and a subsequent plasticizing effect. The increase in water uptake and retention enhanced the capability for controlled protein delivery. Finally, the biocompatibility of the cements was determined; some toxicity of the material during the first hours of culture incubation was observed. Later, toxicity was observed to decrease due to nonreacted monomer leaching, which ensured the low toxicity of the already polymerized phase.
Sigma-Aldrich
Methyl acetoacetate, Arxada quality, ≥99% (GC)
Sigma-Aldrich
L-Glutamine
Sigma-Aldrich
Sodium hydroxide-16O solution, 20 wt. % in H216O, 99.9 atom % 16O
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
Methyl acetoacetate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Methyl acetoacetate, produced by Wacker Chemie AG, Burghausen, Germany, ≥99.0% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
SAFC
L-Glutamine
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Ethanesulfonic acid, 95%
Sigma-Aldrich
Methacrylic acid, contains 250 ppm MEHQ as inhibitor, 99%
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Supelco
Methanol solution, contains 0.10 % (v/v) formic acid, UHPLC, suitable for mass spectrometry (MS), ≥99.5%
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, Vetec, reagent grade, 99%
Sigma-Aldrich
Methyl methacrylate, 99%, stabilized
Sigma-Aldrich
N,N′-Disuccinimidyl carbonate, ≥95%
Sigma-Aldrich
Luperox® AFR40, Benzoyl peroxide solution, 40 wt. % in dibutyl phthalate
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Luperox® A75FP, Benzoyl peroxide, 75% remainder water, contains 25 wt. % water as stabilizer, 75%
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%