Merck
  • FHL1 inhibits the growth of tongue squamous cell carcinoma cells via G1/S cell cycle arrest.
FHL1 inhibits the growth of tongue squamous cell carcinoma cells via G1/S cell cycle arrest.
Molecular medicine reports (2015-05-29)
Wei Ren, Panfeng Lian, Long Cheng, Peiyun Du, Xin Guan, Hongyuan Wang, Lihua Ding, Zhenyang Gao, Xin Huang, Fengjun Xiao, Lisheng Wang, Xiaolin Bi, Qinong Ye, Enqun Wang
ABSTRACT
Four and a half LIM protein 1 (FHL1) has been characterized as a tumor suppressor in various types of tumor. However, the biological function and underlying mechanism of FHL1 in tongue squamous cell carcinoma (TSCC) remain to be elucidated. The present study demonstrated that FHL1 inhibits anchorage‑dependent and ‑independent growth of TSCC cells in vitro and tumor growth in nude mice, as determined by cell proliferation and soft agar assays. Knockdown of FHL1 with FHL1 small interfering RNA (siRNA) promoted tumor growth in nude mice. Mechanistically, flow cytometric analysis showed that knockdown of FHL1 promoted G1/S cell cycle progression. Furthermore, expression of cell cycle‑associated regulators, cyclin D and cyclin E, were detected by western blotting and reverse transcription‑quantitative polymerase chain reaction. Cyclin D and cyclin E were markedly elevated at both the protein and mRNA level in the FHL1 siRNA‑transfected cells. These results suggested that FHL1 has a tumor suppressive role in TSCC and that FHL1 may be a useful target for TSCC gene therapy.
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Cfh
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Fhl1
Sigma-Aldrich
MISSION® esiRNA, targeting human CCNE1
Sigma-Aldrich
MISSION® esiRNA, targeting human GAPDH
Sigma-Aldrich
MISSION® esiRNA, targeting human PCNA
Sigma-Aldrich
PCNA human, recombinant, expressed in baculovirus infected Sf9 cells, ≥90% (SDS-PAGE), ≥90% (HPLC)
Sigma-Aldrich
MISSION® esiRNA, targeting human CFH
Sigma-Aldrich
MISSION® esiRNA, targeting human FHL1