Merck
  • Bisdemethoxycurcumin (BDMC) Alters Gene Expression-associated Cell Cycle, Cell Migration and Invasion and Tumor Progression in Human Lung Cancer NCI-H460 Cells.
Bisdemethoxycurcumin (BDMC) Alters Gene Expression-associated Cell Cycle, Cell Migration and Invasion and Tumor Progression in Human Lung Cancer NCI-H460 Cells.
In vivo (Athens, Greece) (2015-11-08)
Chien-Chih Yu, Mei-Due Yang, Hui-Yi Lin, An-Cheng Huang, Jing-Pin Lin, Chao-Lin Kuo, Kuo-Ching Liu, Hsin-Chung Liu, Su-Tso Yang, Jing-Gung Chung
ABSTRACT
Lung cancer is one of the most common malignancies and a predominant cause of cancer-related death. It can metastasize in almost all organs, and currently, while new cases are increasing, treatment is still insufficient. Bisdemethoxycurcumin (BDMC), one of the components of turmeric, has been known to possess biological activities. However, the effects of BDMC on the genetic level remain unclear. Human lung cancer NCI-H460 cells were treated with 35 μM BDMC for 24 h and cells were harvested for total RNA extraction. The purified RNA was used for cDNA synthesis, labeling, microarray hybridization, and flour-labeled cDNA on-chip hybridization. The expression Console software (Affymetrix) with default RNA parameters was used to detect and quantitate concentrations of fluorescent molecules. The key genes involved and their possible interaction pathways were analyzed by the GeneGo software. Seven genes, such as CCNE2 (cyclin E), associated with cell cycle, were over 4-fold overexpressed, 22 genes, such as ERCC6L (excision repair cross-complementing rodent repair deficiency, complementation group 6-like) associated with DNA damage and repair, were from 3- to 4-fold overexpressed and 266, such as cell division cycle, S-phase associated kinase and associated with cell death, genes were from 2- to 3-fold overexpressed. BDMC induced changes in gene expression that may reveal cytotoxic information on the genetic level while presenting novel biomarkers or targets for treatment of human lung cancer in the future.
Sigma-Aldrich
4-Bromo-3,5-dimethylphenyl N-methylcarbamate, 98%
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, ≥99.5% (GC)
SAFC
L-Glutamine
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
Dimethyl sulfoxide solution, 50 wt. % in H2O
Sigma-Aldrich
L-Glutamine
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., dried, ≤0.02% water
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, Vetec, reagent grade, 99%
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., ACS reagent, ≥99.9% (GC)