Merck
  • Investigation of cellular responses upon interaction with silver nanoparticles.
Investigation of cellular responses upon interaction with silver nanoparticles.
International journal of nanomedicine (2015-09-09)
Ramesh Subbiah, Seong Beom Jeon, Kwideok Park, Sang Jung Ahn, Kyusik Yun
ABSTRACT
In order for nanoparticles (NPs) to be applied in the biomedical field, a thorough investigation of their interactions with biological systems is required. Although this is a growing area of research, there is a paucity of comprehensive data in cell-based studies. To address this, we analyzed the physicomechanical responses of human alveolar epithelial cells (A549), mouse fibroblasts (NIH3T3), and human bone marrow stromal cells (HS-5), following their interaction with silver nanoparticles (AgNPs). When compared with kanamycin, AgNPs exhibited moderate antibacterial activity. Cell viability ranged from ≤ 80% at a high AgNPs dose (40 µg/mL) to >95% at a low dose (10 µg/mL). We also used atomic force microscopy-coupled force spectroscopy to evaluate the biophysical and biomechanical properties of cells. This revealed that AgNPs treatment increased the surface roughness (P<0.001) and stiffness (P<0.001) of cells. Certain cellular changes are likely due to interaction of the AgNPs with the cell surface. The degree to which cellular morphology was altered directly proportional to the level of AgNP-induced cytotoxicity. Together, these data suggest that atomic force microscopy can be used as a potential tool to develop a biomechanics-based biomarker for the evaluation of NP-dependent cytotoxicity and cytopathology.
Sigma-Aldrich
VenPure® SF, powder
Sigma-Aldrich
Silver nitrate, BioXtra, >99% (titration)
Sigma-Aldrich
Silver nitrate, BioReagent, suitable for plant cell culture, >99% (titration)
Sigma-Aldrich
(Tyr[SO3H]27)Cholecystokinin fragment 26-33 Amide, ≥97% (HPLC), powder
Sigma-Aldrich
Sodium borohydride solution, ~12 wt. % in 14 M NaOH
Sigma-Aldrich
Silver nitrate, 99.9999% trace metals basis
Sigma-Aldrich
Sodium borohydride solution, 2.0 M in triethylene glycol dimethyl ether
Sigma-Aldrich
Silver nitrate, anhydrous, ≥99.999% trace metals basis
Sigma-Aldrich
Silver nitrate, ReagentPlus®, ≥99.0% (titration)
Sigma-Aldrich
Silver nitrate, meets analytical specification of Ph. Eur., BP, USP, 99.8-100.5%
Sigma-Aldrich
Silver nitrate, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8%
Sigma-Aldrich
Silver nitrate, puriss. p.a., ≥99.5% (AT)
Sigma-Aldrich
Silver nitrate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium borohydride, purum p.a., ≥96% (gas-volumetric)
Sigma-Aldrich
Sodium borohydride, powder, ≥98.0%
Sigma-Aldrich
Sodium borohydride, granular, 10-40 mesh, 98%
Sigma-Aldrich
Silver nitrate on silica gel, extent of labeling: ~10 wt. % loading, +230 mesh
Sigma-Aldrich
Sodium borohydride, ReagentPlus®, 99%
Sigma-Aldrich
Sodium borohydride, granular, 99.99% trace metals basis
Sigma-Aldrich
Silver nitrate solution, 2.5 % (w/v) AgNO3 in H2O
Sigma-Aldrich
Sodium borohydride, caplets (18 × 10 × 8 mm), 98%