Merck
  • Highly charged cellulose-based nanocrystals as flocculants for harvesting Chlorella vulgaris.
Highly charged cellulose-based nanocrystals as flocculants for harvesting Chlorella vulgaris.
Bioresource technology (2015-07-27)
Dries Vandamme, Samuel Eyley, Guy Van den Mooter, Koenraad Muylaert, Wim Thielemans
ABSTRACT
This study presents a novel flocculant for harvesting Chlorella vulgaris as model species for freshwater microalgae based on cellulose nanocrystals (CNCs), thus synthesized from a renewable and biodegradable resource. Cationic pyridinium groups were grafted onto CNCs by two separate one-pot simultaneous esterification and nucleophilic substitution reactions. Both types of modified CNCs were positively charged in the pH range 4-11. Both reactions yielded CNCs with a high degree of substitution (up to 0.38). A maximum flocculation efficiency of 100% was achieved at a dosage of 0.1 g g(-1) biomass. In contrast to conventional polymer flocculants, cationic CNCs were relatively insensitive to inhibition of flocculation by algal organic matter. The present results highlight the potential of these new type of nanocellulose-based flocculants for microalgae harvesting.
Product Description
Supelco
Dichloromethane solution, contains 10 % (v/v) methanol
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
p-Toluenesulfonyl chloride, ReagentPlus®, ≥99%
Sigma-Aldrich
4-(Bromomethyl)benzoic acid, 97%
Sigma-Aldrich
Pyridine, anhydrous, 99.8%
Sigma-Aldrich
Dichloromethane, anhydrous, ≥99.8%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Dichloromethane, suitable for HPLC, ≥99.9%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
p-Toluenesulfonyl chloride, reagent grade, ≥98%
Sigma-Aldrich
Pyridine, ≥99%
Supelco
Ethanol standards 10% (v/v), 10 % (v/v) in H2O, analytical standard
Sigma-Aldrich
Sulfuric acid, 99.999%