Merck
  • A novel two-dimensional liquid-chromatography method for online prediction of the toxicity of transformation products of benzophenones after water chlorination.
A novel two-dimensional liquid-chromatography method for online prediction of the toxicity of transformation products of benzophenones after water chlorination.
Analytical and bioanalytical chemistry (2015-06-07)
Jian Li, Li-Yun Ma, Li Xu, Zhi-Guo Shi
ABSTRACT
Benzophenone-type UV filters (BPs) are ubiquitous in the environment. Transformation products (TPs) of BPs with suspected toxicity are likely to be produced during disinfection of water by chlorination. To quickly predict the toxicity of TPs, in this study, a novel two-dimensional liquid-chromatography (2D-LC) method was established in which the objective of the first dimension was to separate the multiple components of the BPs sample after chlorination, using a reversed-phase liquid-chromatography mode. A biochromatographic system, i.e. bio-partitioning micellar chromatography with the polyoxyethylene (23) lauryl ether aqueous solution as the mobile phase, served as the second dimension to predict the toxicity of the fraction from the first dimension on the basis of the quantitative retention-activity relationships (QRARs) model. Six BPs, namely 2,4-dihydroxybenzophenone, oxybenzone, 4-hydroxybenzophenone, 2-hydroxy-4-methoxybenzophenone-5-sulfonic acid, 2,2'-dihydroxy-4,4'-dimethoxybenzophenone and 2,2'-dihydroxy-4-methoxybenzophenone, were the target analytes subjected to chlorination. The products of these BPs after chlorination were directly injected to the 2D-LC system for analysis. The results indicated that most TPs may be less toxic than their parent chemicals, but some may be more toxic, and that intestinal toxicity of TPs may be more obvious than blood toxicity. The proposed method is time-saving, high-throughput, and reliable, and has great potential for predicting toxicity or bioactivity of unknown and/or known components in a complex sample. Graphical Abstract The scheme for the 2D-LC online prediction of toxicity of the transformation products of benzophenone-type UV filters after chlorination.
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Acetic acid, for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Ammonium acetate, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
2,2′-Dihydroxy-4-methoxybenzophenone, 98%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium Thiosulfate Solution, 2 g/dL in deionized water
Sigma-Aldrich
Ammonium acetate solution, for molecular biology, 7.5 M
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Ammonium acetate, for molecular biology, ≥98%
Sigma-Aldrich
Ammonium acetate, reagent grade, ≥98%
Sigma-Aldrich
Ammonium acetate, BioXtra, ≥98%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
SAFC
Sodium chloride solution, 5 M