Merck
  • Increased in vitro and in vivo sensitivity of BRCA2-associated pancreatic cancer to the poly(ADP-ribose) polymerase-1/2 inhibitor BMN 673.
Increased in vitro and in vivo sensitivity of BRCA2-associated pancreatic cancer to the poly(ADP-ribose) polymerase-1/2 inhibitor BMN 673.
Cancer letters (2015-04-14)
Alexandra-Zoe Andrei, Anita Hall, Alyssa L Smith, Claire Bascuñana, Abba Malina, Ashton Connor, Gulbeyaz Altinel-Omeroglu, Sidong Huang, Jerry Pelletier, David Huntsman, Steven Gallinger, Atilla Omeroglu, Peter Metrakos, George Zogopoulos
ABSTRACT
BRCA2-associated pancreatic ductal adenocarcinoma (PDAC) may be sensitive to agents that target homology-directed DNA repair, such as DNA crosslinking agents (DCLs) and PARP inhibitors (PARPis). Here, we assessed the sensitivities of BRCA2-deficient (Capan-1) and BRCA2-proficient (MIA PaCa-2) PDAC cell lines to a panel of DCLs and PARPis. Compared to MIA PaCa-2, Capan-1 was significantly more sensitive to all tested DCLs and PARPis, with similar increased sensitivities to cisplatin and the PARPi BMN 673 compared to other DCLs and the PARPi veliparib. We provide further support for this observation by showing that shRNA-mediated BRCA2 knockdown in PANC-1, a BRCA2-proficient cell line, induces sensitization to cisplatin and BMN 673 but not to veliparib. These findings were validated in a PDAC murine xenograft model derived from a patient with bi-allelic BRCA2 mutations. We found 64% and 61% tumor growth inhibition of this xenograft with cisplatin and BMN 673 treatments, respectively. Cisplatin and BMN 673 treatments reduced cellular proliferation and induced apoptosis. Our findings support a personalized treatment approach for BRCA2-associated PDAC.
Sigma-Aldrich
cis-Diamineplatinum(II) dichloride, ≥99.9% trace metals basis
SAFC
N,N-Dimethylacetamide, Ph. Eur.
Sigma-Aldrich
Dimethyl sulfoxide, Vetec, reagent grade, 99%
Sigma-Aldrich
N,N-Dimethylacetamide, suitable for peptide synthesis, ≥99.8% (GC)
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
N,N-Dimethylacetamide-d9, 99 atom % D
Sigma-Aldrich
N,N-Dimethylacetamide, ReagentPlus®, ≥99%
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Oxaliplatin, powder
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
N,N-Dimethylacetamide, anhydrous, 99.8%
Sigma-Aldrich
Carboplatin
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
trans-Platinum(II)diammine dichloride
Sigma-Aldrich
cis-Diammineplatinum(II) dichloride, crystalline
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%