Merck
  • Coordinated loss of microRNA group causes defenseless signaling in malignant lymphoma.
Coordinated loss of microRNA group causes defenseless signaling in malignant lymphoma.
Scientific reports (2015-12-08)
Makoto Yamagishi, Harutaka Katano, Tsunekazu Hishima, Tatsu Shimoyama, Yasunori Ota, Kazumi Nakano, Takaomi Ishida, Seiji Okada, Toshiki Watanabe
ABSTRACT
Biological robustness is exposed to stochastic perturbations, which should be controlled by intrinsic mechanisms; the promiscuous signaling network without appropriate alleviation is the true nature of cancer cells. B cell receptor (BCR) signaling is a major source of gene expression signature important for B cell. It is still unclear the mechanism by which the expression of functionally important genes is continuously deregulated in malignant lymphomas. Using RISC-capture assay, we reveal that multiple BCR signaling factors are persistently regulated by microRNA (miRNA) in human B cells. Clinical samples from patients with diffuse large B-cell lymphoma (DLBCL, n = 83) show loss of an essential miRNA set (miR-200c, miR-203, miR-31). Conventional screening and RISC profiling identify multiple targets (CD79B, SYK, PKCβII, PLCγ1, IKKβ, NIK, MYD88, PI3K class I (α/β/δ/γ), RasGRP3); signaling network habitually faces interference composed by miRNA group in normal B cells. We demonstrate that simultaneous depletion of the key miRNAs enhances translation of the multiple targets and causes chronic activation of NF-κB, PI3K-Akt, and Ras-Erk cascades, leading to B cell transformation. This study suggests that compensatory actions by multiple miRNAs rather than by a single miRNA ensure robustness of biological processes.
Sigma-Aldrich
Trichloroacetic acid, ACS reagent, for the determination of Fe in blood according to Heilmeyer, ≥99.5%
Sigma-Aldrich
Trichloroacetic acid, BioUltra, ≥99.5% (T)
Sigma-Aldrich
Sodium trichloroacetate, 97%
Sigma-Aldrich
Trichloroacetic acid, suitable for electrophoresis, suitable for fixing solution (for IEF and PAGE gels), ≥99%
Sigma-Aldrich
Trichloroacetic acid, ≥99.0% (titration)
Sigma-Aldrich
Trichloroacetic acid, ACS reagent, ≥99.0%
Sigma-Aldrich
Nitrotetrazolium Blue chloride, powder, electrophoresis grade
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Sigma-Aldrich
Trichloroacetic acid, BioXtra, ≥99.0%
Sigma-Aldrich
Nitrotetrazolium Blue chloride, ≥90.0% (HPLC)
Supelco
DL-Dithiothreitol solution, 1 M in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Trichloroacetic acid solution, 6.1 N
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, BioUltra, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Z-Leu-Leu-Leu-al, ≥90% (HPLC)
Sigma-Aldrich
IgG from mouse serum, reagent grade, ≥95% (SDS-PAGE), lyophilized powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, Vetec, reagent grade, 98%