Merck
  • Heterogeneous Nuclear Ribonucleoprotein A1 Improves the Intestinal Injury by Regulating Apoptosis Through Trefoil Factor 2 in Mice with Anti-CD3-induced Enteritis.
Heterogeneous Nuclear Ribonucleoprotein A1 Improves the Intestinal Injury by Regulating Apoptosis Through Trefoil Factor 2 in Mice with Anti-CD3-induced Enteritis.
Inflammatory bowel diseases (2015-04-23)
Katsuyoshi Ando, Mikihiro Fujiya, Hiroaki Konishi, Nobuhiro Ueno, Yuhei Inaba, Kentaro Moriichi, Katsuya Ikuta, Hiroki Tanabe, Takaaki Ohtake, Yutaka Kohgo
ABSTRACT
The role of hnRNP A1 in the onset of intestinal inflammation remains unclear. This study investigated the function of hnRNP A1 in mice enteritis models. C57Bl6/J mice were intraperitoneally injected with anti-CD3 antibodies to develop enteritis. In the DSS-induced colitis group, the mice were allowed free access to 3% DSS solution in their drinking water for 5 days. 3H-mannitol flux and complementary DNA array tests were used to assess the intestinal barrier function and messenger RNA (mRNA) expression, respectively. Real-time PCR was performed after immunoprecipitation with anti-hnRNP antibodies to determine the specific mRNA binding of hnRNP A1. The hnRNP A1 expression was increased in the intestine of the mouse at 24 hours after treatment with anti-CD3 antibodies and 5 days after starting DSS administration. Small interfering RNA (siRNA) against hnRNP A1 exacerbated the intestinal injuries in both models. According to the microarray analysis, trefoil factor 2 (TFF2) was identified as a candidate molecule targeted by hnRNP A1 in the anti-CD3 antibody-induced enteritis group. Moreover, the binding between hnRNP A1 and TFF2 mRNA significantly increased in the enteritis mice, and the administration of siRNA against either hnRNP A1 or TFF2 exacerbated the degree of intestinal injury. In the DSS-induced colitis group, treatment with the siRNA of hnRNP A1 worsened the intestinal injury, while the expression of TFF3 did not change. hnRNP A1 improves intestinal injury in anti-CD3 antibody-induced enteritis mice through the upregulation of TFF2, which regulates apoptosis and enhances epithelial restoration, whereas this molecule ameliorates DSS-induced colitis through a different pathway.
Sigma-Aldrich
Chloroform, ACS reagent, ≥99.8%, contains amylenes as stabilizer
Sigma-Aldrich
Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, ACS reagent, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Supelco
Chloroform, suitable for HPLC, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, ≥99%, PCR Reagent, contains amylenes as stabilizer
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Sigma-Aldrich
L-Glutamine
Sigma-Aldrich
o-Xylene, anhydrous, 97%
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
SAFC
L-Glutamine
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Supelco
Ethanol standards 10% (v/v), 10 % (v/v) in H2O, analytical standard
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, BioUltra, ≥99% (titration)
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, Vetec, reagent grade, 98%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)