Merck
  • Feline calicivirus and other respiratory pathogens in cats with Feline calicivirus-related symptoms and in clinically healthy cats in Switzerland.
Feline calicivirus and other respiratory pathogens in cats with Feline calicivirus-related symptoms and in clinically healthy cats in Switzerland.
BMC veterinary research (2015-11-15)
Alice Berger, Barbara Willi, Marina L Meli, Felicitas S Boretti, Sonja Hartnack, Anou Dreyfus, Hans Lutz, Regina Hofmann-Lehmann
ABSTRACT
Cats with feline calicivirus (FCV)-related symptoms are commonly presented to veterinary practitioners. Various clinical manifestations have been attributed to FCV, i.e. upper respiratory tract disease (URTD), oral ulcerations, gingivostomatitis, limping syndrome and virulent systemic disease. Additionally, healthy cats can shed FCV. The aims of this study were 1) to investigate the frequency of FCV in cats with FCV-related symptoms and in healthy cats in Switzerland, 2) to assess risk and protective factors for infection, such as signalment, housing conditions, vaccination, and co-infection with URTD-associated pathogens, and 3) to address the association between clinical symptoms and FCV infection. Oropharyngeal, nasal and conjunctival swabs were collected in 24 veterinary practices from 200 FCV-suspect and 100 healthy cats originating from 19 cantons of Switzerland. The samples were tested for FCV using virus isolation and reverse-transcription real-time quantitative polymerase chain reaction (qPCR) and for feline herpesvirus-1 (FHV-1), Mycoplasma felis, Chlamydophila felis, Bordetella bronchiseptica using real-time qPCR. Within the two populations (FCV-suspect/healthy), the observed PCR prevalences were: FCV 45%/8%, FHV-1 20%/9%, C. felis 8%/1%, B. bronchiseptica 4%/2%, M. felis 47%/31% and any co-infections thereof 40%/14%. Based on multivariable regression models amongst FCV-suspect cats (odds ratio [95% confidence interval]), co-infection with M. felis (1.75 [0.97; 3.14]), group housing (2.11 [1.02; 4.34]) and intact reproductive status (1.80 [0.99; 3.28]) were found to be risk factors for FCV infection. In healthy cats, intact reproductive status (22.2 [1.85; 266.7]) and group housing (46.4 [5.70; 377.7]) were found to be associated with FCV infection. Based on an univariable approach, FCV-suspect cats were found to be significantly less often FCV-positive when vaccinated (0.48 [0.24; 0.94]). Oral ulcerations, salivation, gingivitis and stomatitis, but not classical signs of URTD were significantly associated with FCV infection (all p < 0.001). FCV was detected in less than half of the cats that were judged FCV-suspect by veterinary practitioners. For a clinical diagnosis, FCV-related symptoms should be revisited. FCV infection was present in some healthy cats, underlining the importance of asymptomatic carriers in FCV epidemiology. To reduce FCV-related problems in multi-cat environments, reduction of group size in addition to the generally recommended vaccination are advocated.
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, for molecular biology, 10 M in H2O
Sigma-Aldrich
Sodium hydroxide, BioUltra, for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium hydroxide, anhydrous, free-flowing, Redi-Dri, reagent grade, ≥98%, pellets
Sigma-Aldrich
Sodium hydroxide, ultra dry, powder or crystals, 99.99% trace metals basis
Sigma-Aldrich
Sodium hydroxide, reagent grade, ≥98%, pellets (anhydrous)
Sigma-Aldrich
Sodium hydroxide solution, purum, ≥32%
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, powder
Sigma-Aldrich
Sodium hydroxide, puriss., meets analytical specification of Ph. Eur., BP, NF, E524, 98-100.5%, pellets
Sigma-Aldrich
Sodium hydroxide, puriss. p.a., ACS reagent, K ≤0.02%, ≥98.0% (T), pellets
Sigma-Aldrich
Sodium hydroxide, puriss. p.a., ACS reagent, reag. Ph. Eur., K ≤0.02%, ≥98%, pellets
Sigma-Aldrich
Sodium hydroxide, ACS reagent, ≥97.0%, pellets
Sigma-Aldrich
Sodium hydroxide, pellets, semiconductor grade, 99.99% trace metals basis
Sigma-Aldrich
Sodium hydroxide, BioXtra, ≥98% (acidimetric), pellets (anhydrous)
Sigma-Aldrich
Sodium hydroxide solution, 5.0 M
Sigma-Aldrich
Sodium hydroxide, beads, 16-60 mesh, reagent grade, 97%
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, flakes
Sigma-Aldrich
Sodium hydroxide solution, 50% in H2O
Sigma-Aldrich
L-Glutamine
Sigma-Aldrich
Sodium hydroxide-16O solution, 20 wt. % in H216O, 99.9 atom % 16O
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
SAFC
L-Glutamine
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source