Merck
  • Early post mortem expression of genes related to tenderization in two Italian Simmental young bulls' skeletal muscles differing in contractile type.
Early post mortem expression of genes related to tenderization in two Italian Simmental young bulls' skeletal muscles differing in contractile type.
Animal science journal = Nihon chikusan Gakkaiho (2015-07-15)
Elena Saccà, Mirco Corazzin, Nicoletta Pizzutti, Giovanna Lippe, Edi Piasentier
ABSTRACT
The early post mortem expression of eight genes potentially involved in meat ageing process and the tenderness of two Italian Simmental young bulls' (Bos taurus) skeletal muscles differing in their contractile type were evaluated. Samples of Longissimus lumborum (LL) and Infraspinatus (IS) muscles were collected from 17 bulls. The messenger RNA (mRNA) abundances of calpain-1, calpain-2, calpastatin, caspase 3, caspase 9, heat shock protein 27 (Hsp27), Hsp40 and Hsp70 were detected by quantitative PCR. The myosin heavy chain-slow and -fast isoform content, the pH48h and the lipid content of the muscles were in line with the contractile and metabolic type. In comparison with the fast LL, the slow IS showed a lower calpain-1/calpastatin mRNA content ratio after slaughtering and a higher Warner-Bratzler Initial Yield value after 7 days of ageing. Hsp27 and Hsp70 mRNA abundances were significantly lower in LL than IS, highlighting their potential role in the ageing process of bovine muscles.
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Monoclonal Anti-Myosin (Skeletal, Slow) antibody produced in mouse, clone NOQ7.5.4D, ascites fluid
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Anti-Actin antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
Anti-Myosin (Skeletal, Fast) antibody, Mouse monoclonal, clone MY-32, purified from hybridoma cell culture
Supelco
Methanol solution, contains 0.10 % (v/v) formic acid, UHPLC, suitable for mass spectrometry (MS), ≥99.5%
Sigma-Aldrich
Hydriodic acid, 57 wt. %, distilled, 99.999% trace metals basis
Sigma-Aldrich
Hydriodic acid, contains ≤1.5% hypophosphorous acid as stabilizer, ACS reagent, ≥47.0%
Sigma-Aldrich
Hydriodic acid, contains no stabilizer, distilled, 57 wt. % in H2O, 99.99% trace metals basis
Sigma-Aldrich
Hydriodic acid, 57 wt. % in H2O, distilled, stabilized, 99.95%
Sigma-Aldrich
Hydriodic acid, contains no stabilizer, ACS reagent, 55%