Merck
  • Comparative Two-Dimensional Gel Electrophoresis of Trypanosoma cruzi Mammalian-Stage Forms in an Alkaline pH Range.
Comparative Two-Dimensional Gel Electrophoresis of Trypanosoma cruzi Mammalian-Stage Forms in an Alkaline pH Range.
Protein and peptide letters (2015-09-16)
Adriana D Magalhães, Rayner Myr L Queiroz, Izabela M D Bastos, Jaime M Santana, Marcelo V Sousa, Carlos André O Ricart, Sébastien Charneau
ABSTRACT
It is estimated that several million people are currently infected worldwide by the protozoan parasite, Trypanosoma cruzi, which causes Chagas disease. After mammalian host infection, a fundamental event is the differentiation from infective trypomastigotes into replicative amastigotes (amastigogenesis) inside host-cells. To unravel the particularities of both forms, it is essential to identify molecules presented in each form. Since T. cruzi gene expression regulation occurs mainly at posttranscriptional level, a proteomic approach is appropriate. Due to intrinsic difficulties with performing 2-DE in the alkaline pH range, there are no reports on 2-DE-based comparative proteome analysis of T. cruzi mammalianstage forms that focus on alkaline polypeptides. Here, we performed a comparative proteome analysis between tissue culture- derived trypomastigotes and extracellular amastigote-like cells using conditions optimized for the 6-11 pH range followed by identification by MALDI-TOF/TOF technology. The alkaline 2-DE maps from both forms show that proteins with a pI above 7.0 were not underrepresented (= 65% of proteins detected). Moreover the differences in protein expression between the Human-hosted T. cruzi forms corroborated previous proteomic studies and corresponded to their biological traits.
Sigma-Aldrich
Urea-12C, 99.9 atom % 12C
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
Thiourea, ACS reagent, ≥99.0%
Sigma-Aldrich
Urea solution, 40 % (w/v) in H2O
Sigma-Aldrich
Thiourea, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Urea solution, BioUltra, ~8 M in H2O
Supelco
DL-Dithiothreitol solution, 1 M in H2O
Supelco
Urea, 8 M (after reconstitution with 16 mL high purity water)
Sigma-Aldrich
Urea, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%, 99.0-101.0% (calc. on dry substance)
Sigma-Aldrich
Urea, BioUltra, for molecular biology, 99% (T)
Sigma-Aldrich
Urea, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99%
Sigma-Aldrich
Urea, BioXtra, pH 7.5-9.5 (20 °C, 5 M in H2O)
Sigma-Aldrich
Urea, ReagentPlus®, ≥99.5%, pellets
Sigma-Aldrich
Urea, powder, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Urea, ACS reagent, 99.0-100.5%
Sigma-Aldrich
Urea, suitable for electrophoresis
Sigma-Aldrich
Urea, meets USP testing specifications