Merck
  • Proteomic screening and identification of microRNA-128 targets in glioma cells.
Proteomic screening and identification of microRNA-128 targets in glioma cells.
Proteomics (2015-03-18)
Bin Yang, Shan Wang, Jiawei Zeng, Yu Zhang, Xiangbin Ruan, Wei Han, Bin Yin, Jiangang Yuan, Boqin Qiang, Wantao Ying, Xiaohong Qian, Xiaozhong Peng
ABSTRACT
Brain-enriched miR-128 is repressed in glioma cells, and could inhibit the proliferation of gliomas by targeting genes such as E2F3a and BMI1. To identify more targets of miR-128 in glioblastoma multiforme, the pulse stable isotope labeling with amino acids in cell culture (pSILAC) technique was used to test its impact on whole protein synthesis in T98G glioma cells. We successfully identified 1897 proteins, of which 1459 proteins were quantified. Among them, 133 proteins were downregulated after the overexpression of miR-128. Through predictions using various bioinformatics tools, 13 candidate target genes were chosen. A luciferase assay validated that 11 of 13 selected genes were potential targets of miR-128, and a mutagenesis experiment confirmed CBFB, CORO1C, GLTP, HnRNPF, and TROVE2 as the target genes. Moreover, we observed that the expression of CORO1C, TROVE2, and HnRNPF were higher in glioma cell lines compared to normal brain tissues and presented a tendency toward downregulation after overexpression of miR-128 in T98G cells. Furthermore, we have validated that CORO1C, TROVE2, and HnRNPF could inhibit glioma cell proliferation. In sum, our data showed that the integration of pSILAC and bioinformatics analysis was an efficient method for seeking the targets of miRNAs, and plentiful targets of miR-128 were screened and laid the foundation for research into the miR-128 regulation network.
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Anti-TROVE2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC
Sigma-Aldrich
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Citrate Concentrated Solution, BioUltra, for molecular biology, 1 M in H2O
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, for molecular biology
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride solution, 5 M