Merck
  • In vitro growth versus inhibition of growth of Malassezia pachydermatis in the presence of the antibacterial drug gentamicin.
In vitro growth versus inhibition of growth of Malassezia pachydermatis in the presence of the antibacterial drug gentamicin.
Journal of medical microbiology (2015-01-30)
Elisabetta Buommino, Adone Baroni, Annamaria Parisi, Flavia Elena Borriello, Pina Caputo, Giovanna Donnarumma, Sandra Nizza, Francesca Paola Nocera, Filomena Fiorito, Luisa De Martino
ABSTRACT
Malassezia pachydermatis is part of the normal cutaneous microbiota of most warm-blooded vertebrates and is associated with otitis externa and seborrhoeic dermatitis in dogs and cats. In this study, we evaluated the growth capacity of nine M. pachydermatis strains on Sabouraud medium in the presence of a high concentration of gentamicin. Strains of M. pachydermatis cultured on Sabouraud medium in the presence of 50 and 100 µg gentamicin ml(-1) displayed different growth patterns such as growth or lack of growth. We hypothesized that this difference in growth of M. pachydermatis strains was correlated with the different genotypes of the strains. Random amplification of polymorphic DNA (RAPD) was applied for genetic typing of M. pachydermatis isolates, derived from the external ears of house pet cats suffering from otitis externa. The M. pachydermatis strains were cultured on commercial or home-made Sabouraud medium supplemented or not with gentamicin. RAPD analysis demonstrated a genetic heterogeneity between each strain. In particular, five out of nine strains tested were able to form colonies in the presence of gentamicin. However, a correlation between M. pachydermatis genotype and growth capacity in the presence of gentamicin was not widely demonstrated.
Sigma-Aldrich
D-(+)-Glucose, Vetec, reagent grade, ≥99.5% (HPLC)
Sigma-Aldrich
Sodium acetate, Vetec, reagent grade, 98%
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium acetate, powder, BioReagent, suitable for electrophoresis, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
Sodium acetate, anhydrous, for molecular biology, ≥99%
Sigma-Aldrich
D-(+)-Glucose, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.5%
Sigma-Aldrich
Sodium acetate, meets USP testing specifications, anhydrous
Sigma-Aldrich
Sodium acetate, BioXtra, ≥99.0%
Sigma-Aldrich
D-(+)-Glucose, ACS reagent
Sigma-Aldrich
D-(+)-Glucose, Hybri-Max, powder, BioReagent, suitable for hybridoma
Sigma-Aldrich
D-(+)-Glucose, suitable for mouse embryo cell culture, ≥99.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Chloroform, ≥99%, PCR Reagent, contains amylenes as stabilizer
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium acetate, 99.995% trace metals basis
Sigma-Aldrich
Sodium acetate, anhydrous, BioUltra, for luminescence, for molecular biology, ≥99.0% (NT)
Sigma-Aldrich
D-(+)-Glucose, BioUltra, anhydrous, ≥99.5% (sum of enantiomers, HPLC)
Sigma-Aldrich
Chloroform, ACS reagent, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, ACS reagent, ≥99.8%, contains amylenes as stabilizer
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Supelco
Chloroform, suitable for HPLC, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
D-Glucose-12C6, 16O6, 99.9 atom % 16O, 99.9 atom % 12C
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium acetate solution, BioUltra, for molecular biology, ~3 M in H2O
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate