Merck
  • G-quadruplex enhanced fluorescence of DNA-silver nanoclusters and their application in bioimaging.
G-quadruplex enhanced fluorescence of DNA-silver nanoclusters and their application in bioimaging.
Nanoscale (2015-07-18)
Jinbo Zhu, Libing Zhang, Ye Teng, Baohua Lou, Xiaofang Jia, Xiaoxiao Gu, Erkang Wang
ABSTRACT
Guanine proximity based fluorescence enhanced DNA-templated silver nanoclusters (AgNCs) have been reported and applied for bioanalysis. Herein, we studied the G-quadruplex enhanced fluorescence of DNA-AgNCs and gained several significant conclusions, which will be helpful for the design of future probes. Our results demonstrate that a G-quadruplex can also effectively stimulate the fluorescence potential of AgNCs. The major contribution of the G-quadruplex is to provide guanine bases, and its special structure has no measurable impact. The DNA-templated AgNCs were further analysed by native polyacrylamide gel electrophoresis and the guanine proximity enhancement mechanism could be visually verified by this method. Moreover, the fluorescence emission of C3A (CCCA)4 stabilized AgNCs was found to be easily and effectively enhanced by G-quadruplexes, such as T30695, AS1411 and TBA, especially AS1411. Benefiting from the high brightness of AS1411 enhanced DNA-AgNCs and the specific binding affinity of AS1411 for nucleolin, the AS1411 enhanced AgNCs can stain cancer cells for bioimaging.
Sigma-Aldrich
Silver nitrate, anhydrous, ≥99.999% trace metals basis
Sigma-Aldrich
Silver nitrate, BioReagent, suitable for plant cell culture, >99% (titration)
Sigma-Aldrich
Silver nitrate, BioXtra, >99% (titration)
Sigma-Aldrich
Sodium borohydride solution, 2.0 M in triethylene glycol dimethyl ether
Sigma-Aldrich
Sodium borohydride solution, ~12 wt. % in 14 M NaOH
Sigma-Aldrich
Silver nitrate, 99.9999% trace metals basis
Sigma-Aldrich
VenPure® SF, powder
Sigma-Aldrich
Silver nitrate solution, 2.5 % (w/v) AgNO3 in H2O
Sigma-Aldrich
Sodium borohydride, purum p.a., ≥96% (gas-volumetric)
Sigma-Aldrich
Sodium borohydride, ReagentPlus®, 99%
Sigma-Aldrich
Sodium borohydride, granular, 10-40 mesh, 98%
Sigma-Aldrich
Sodium borohydride, caplets (18 × 10 × 8 mm), 98%
Sigma-Aldrich
Sodium borohydride, granular, 99.99% trace metals basis
Sigma-Aldrich
Silver nitrate on silica gel, extent of labeling: ~10 wt. % loading, +230 mesh
Sigma-Aldrich
Sodium borohydride, powder, ≥98.0%