Merck
  • Mammalian target of rapamycin overexpression antagonizes chronic hypoxia-triggered pulmonary arterial hypertension via the autophagic pathway.
Mammalian target of rapamycin overexpression antagonizes chronic hypoxia-triggered pulmonary arterial hypertension via the autophagic pathway.
International journal of molecular medicine (2015-05-29)
Lingxia Li, Xiaochuang Wang, Lina Wang, Li Qu, Xinye Zhu, Manxiang Li, Xiaoyan Dang, Ping Li, Yanxia Gao, Zhuo Peng, Longfei Pan, Li Wan
ABSTRACT
Pulmonary arterial hypertension (PAH) is a progressive pulmonary vascular disorder with high morbidity and mortality, and is characterized by excessive growth of endothelial cells. Recently, the mammalian target of rapamycin (mTOR) has attracted increasing attention due to its potential as a therapeutic target against certain diseases associated with proliferative and metabolic abnormalities. However, the effect on mTOR on PAH has not yet been elucidated. In the present study, a marked downregulation of mTOR was observed in PAH patients. Following construction of a mouse model of PAH by chronic exposure to hypoxia, adenovirus-mediated upregulation of mTOR significantly attenuated right ventricular systolic pressure, right ventricular hypertrophy and wall thickness of pulmonary arterioles, indicating a protective effect of mTOR on PAH. Further analysis confirmed that mTOR overexpression inhibited autophagy triggered by hypoxia through blocking light chain 3 II expression and increasing p62 levels. In vitro, hypoxia enhanced the proliferation of human pulmonary artery endothelial cells (PAECs), which was markedly abrogated by mTOR overexpression. Of note, upregulation of mTOR inhibited the hypoxia-induced autophagy pathway, which contributed to cell proliferation, while silencing of autophagy by RNA interference with ATG5 significantly inhibited cell proliferation. In conclusion, the results of the present study suggested a potential protective effect of mTOR on the progression of PAH by suppressing PAEC proliferation through blocking the autophagic pathway. Therefore, the present study suggested that mTOR is a promising therapeutic agent against PAH.
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
MISSION® esiRNA, targeting human ANXA3
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Atg5
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, 98%
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
MISSION® esiRNA, targeting human MAP1LC3A
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
MISSION® esiRNA, targeting human ATG5
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder