Merck
  • Decreased miR122 in hepatocellular carcinoma leads to chemoresistance with increased arginine.
Decreased miR122 in hepatocellular carcinoma leads to chemoresistance with increased arginine.
Oncotarget (2015-04-01)
Takahiro Kishikawa, Motoyuki Otsuka, Poh Seng Tan, Motoko Ohno, Xiaochen Sun, Takeshi Yoshikawa, Chikako Shibata, Akemi Takata, Kentaro Kojima, Kenji Takehana, Maki Ohishi, Sana Ota, Tomoyuki Noyama, Yuji Kondo, Masaya Sato, Tomoyoshi Soga, Yujin Hoshida, Kazuhiko Koike
ABSTRACT
Reduced expression of microRNA122 (miR122), a liver-specific microRNA, is frequent in hepatocellular carcinoma (HCC). However, its biological significances remain poorly understood. Because deregulated amino acid levels in cancers can affect their biological behavior, we determined the amino acid levels in miR122-silenced mouse liver tissues, in which intracellular arginine levels were significantly increased. The increased intracellular arginine levels were through upregulation of the solute carrier family 7 (SLC7A1), a transporter of arginine and a direct target of miR122. Arginine is the substrate for nitric oxide (NO) synthetase, and intracellular NO levels were increased in miR122-silenced HCC cells, with increased resistance to sorafenib, a multikinase inhibitor. Conversely, maintenance of the miR122-silenced HCC cells in arginine-depleted culture media, as well as overexpression of miR122 in miR122-low-expressing HCC cells, reversed these effects and rendered the cells more sensitive to sorafenib. Using a reporter knock-in construct, chemical compounds were screened, and Wee1 kinase inhibitor was identified as upregulators of miR122 transcription, which increased the sensitivity of the cells to sorafenib. These results provide an insight into sorafenib resistance in miR122-low HCC, and suggest that arginine depletion or a combination of sorafenib with the identified compound may provide promising approaches to managing this HCC subset.
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Urea solution, BioUltra, ~8 M in H2O
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid
Sigma-Aldrich
Urea-12C, 99.9 atom % 12C
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Urea solution, 40 % (w/v) in H2O
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC), free-flowing, Redi-Dri
Supelco
Urea, 8 M (after reconstitution with 16 mL high purity water)
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 20% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 10% in H2O
Sigma-Aldrich
Urea, BioUltra, for molecular biology, 99% (T)
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Urea, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%, 99.0-101.0% (calc. on dry substance)
Sigma-Aldrich
PTIO
Sigma-Aldrich
Urea, suitable for electrophoresis
Sigma-Aldrich
Urea, meets USP testing specifications
Sigma-Aldrich
Urea, BioXtra, pH 7.5-9.5 (20 °C, 5 M in H2O)
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Urea, ReagentPlus®, ≥99.5%, pellets
Sigma-Aldrich
Urea, ACS reagent, 99.0-100.5%
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%