Merck
  • Impairment of antioxidant defense via glutathione depletion sensitizes acute lymphoblastic leukemia cells for Smac mimetic-induced cell death.
Impairment of antioxidant defense via glutathione depletion sensitizes acute lymphoblastic leukemia cells for Smac mimetic-induced cell death.
Oncogene (2014-11-11)
H Schoeneberger, K Belz, B Schenk, S Fulda
ABSTRACT
Evasion of apoptosis in pediatric acute lymphoblastic leukemia (ALL) is linked to aberrant expression of inhibitor of apoptosis (IAP) proteins and dysregulated redox homeostasis, rendering leukemic cells vulnerable to redox-targeting therapies. Here we discover that inhibition of antioxidant defenses via glutathione (GSH) depletion by buthionine sulfoximine (BSO) primes ALL cells for apoptosis induced by the Smac mimetic BV6 that antagonizes IAP proteins. Similarly, BSO cooperates with BV6 to induce cell death in patient-derived primary leukemic samples, underscoring the clinical relevance. In contrast, BSO does not sensitize non-malignant lymphohematopoietic cells from healthy donors toward BV6, pointing to some tumor selectivity. Mechanistically, both agents cooperate to stimulate reactive oxygen species (ROS) production, which is required for BSO/BV6-induced cell death, as ROS inhibitors (that is, N-acetylcysteine, MnTBAP, Trolox) significantly rescue cell death. Further, BSO and BV6 cooperate to trigger lipid peroxidation, which is necessary for cell death, as genetic or pharmacological blockage of lipid peroxidation by GSH peroxidase 4 (GPX4) overexpression or α-tocopherol significantly inhibits BSO/BV6-mediated cell death. Consistently, GPX4 knockdown or GPX4 inhibitor RSL3 enhances lipid peroxidation and cell death by BSO/BV6 cotreatment. The discovery of redox regulation of Smac mimetic-induced cell death has important implications for developing rational Smac mimetic-based combination therapies.
Sigma-Aldrich
MISSION® esiRNA, targeting human RALBP1
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Ralbp1
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Bromobimane, suitable for fluorescence, BioReagent, ≥95% (HPCE)
Sigma-Aldrich
Bromobimane, ≥97% (HPLC)
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
MISSION® esiRNA, targeting human RIPK1
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
MISSION® esiRNA, targeting human UQCRFS1
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Ripk1
Sigma-Aldrich
L-Glutathione reduced, suitable for cell culture, BioReagent, ≥98.0%, powder
Sigma-Aldrich
L-Glutathione reduced, ≥98.0%
Sigma-Aldrich
L-Glutathione reduced, BioXtra, ≥98.0%
Sigma-Aldrich
L-Glutathione reduced, Vetec, reagent grade, ≥98%