Merck
  • Interferon-Gamma Enhances TLR3 Expression and Anti-Viral Activity in Keratinocytes.
Interferon-Gamma Enhances TLR3 Expression and Anti-Viral Activity in Keratinocytes.
The Journal of investigative dermatology (2015-03-31)
A I Kajita, Shin Morizane, Tetsuya Takiguchi, Takenobu Yamamoto, Masao Yamada, Keiji Iwatsuki
ABSTRACT
Toll-like receptors (TLRs) recognize specific microbial products in the innate immune response. TLR3, a double-stranded RNA sensor, is thought to have an important role in viral infections, but the regulation of TLR3 expression and its function in keratinocytes are not fully understood. Here we show the Th1 cytokine IFN-γ increased the TLR3 expression via STAT1 in cultured normal human epidermal keratinocytes (NHEKs). Co-stimulation with IFN-γ and the TLR3 ligand poly (I:C) synergistically increased the expression of IFN-β, IL-6, IL-8, and human β-defensin-2 in NHEKs compared with poly (I:C) or IFN-γ alone. These synergistic inductions were significantly inhibited by an endosomal acidification inhibitor, chloroquine, and by TLR3 siRNA. Co-stimulation with IFN-γ and poly (I:C) also significantly enhanced the anti-viral activity against herpes simplex virus type-1 in NHEKs compared with poly (I:C) or IFN-γ alone. In addition to the in vitro findings, an immunohistochemical analysis revealed IFN-γ-positive cells surrounding herpetic vesicles. These findings indicate that IFN-γ might contribute to the innate immune response to cutaneous viral infections by enhancing TLR3 expression and function in keratinocytes.
Sigma-Aldrich
MISSION® esiRNA, targeting human TLR3
Sigma-Aldrich
Interleukin-6 human, IL-6, recombinant, expressed in HEK 293 cells, suitable for cell culture
Sigma-Aldrich
Interleukin-6 from rat, IL-6, recombinant, expressed in E. coli, buffered aqueous solution, suitable for cell culture
Sigma-Aldrich
Interleukin-6 human, IL-6, recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture
Sigma-Aldrich
Sulfuric acid, 99.999%
Sigma-Aldrich
IL-6 from mouse, Animal-component free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC)
Sigma-Aldrich
Interleukin-6 human, Animal-component free, recombinant, expressed in E. coli, suitable for cell culture
Sigma-Aldrich
Interleukin-6 from mouse, IL-6, recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture, carrier free
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Tlr3
Sigma-Aldrich
IL-6 from rat, recombinant, expressed in E. coli, ≥90% (SDS-PAGE), ≥90% (HPLC)