Merck
  • Bordetella pertussis Adenylate Cyclase Toxin Blocks Induction of Bactericidal Nitric Oxide in Macrophages through cAMP-Dependent Activation of the SHP-1 Phosphatase.
Bordetella pertussis Adenylate Cyclase Toxin Blocks Induction of Bactericidal Nitric Oxide in Macrophages through cAMP-Dependent Activation of the SHP-1 Phosphatase.
Journal of immunology (Baltimore, Md. : 1950) (2015-04-17)
Ondrej Cerny, Jana Kamanova, Jiri Masin, Ilona Bibova, Karolina Skopova, Peter Sebo
ABSTRACT
The adenylate cyclase toxin-hemolysin (CyaA) plays a key role in the virulence of Bordetella pertussis. CyaA penetrates complement receptor 3-expressing phagocytes and catalyzes uncontrolled conversion of cytosolic ATP to the key second messenger molecule cAMP. This paralyzes the capacity of neutrophils and macrophages to kill bacteria by complement-dependent oxidative burst and opsonophagocytic mechanisms. We show that cAMP signaling through the protein kinase A (PKA) pathway activates Src homology domain 2 containing protein tyrosine phosphatase (SHP) 1 and suppresses production of bactericidal NO in macrophage cells. Selective activation of PKA by the cell-permeable analog N(6)-benzoyladenosine-3',5'-cyclic monophosphate interfered with LPS-induced inducible NO synthase (iNOS) expression in RAW264.7 macrophages, whereas inhibition of PKA by H-89 largely restored the production of iNOS in CyaA-treated murine macrophages. CyaA/cAMP signaling induced SHP phosphatase-dependent dephosphorylation of the c-Fos subunit of the transcription factor AP-1 and thereby inhibited TLR4-triggered induction of iNOS gene expression. Selective small interfering RNA knockdown of SHP-1, but not of the SHP-2 phosphatase, rescued production of TLR-inducible NO in toxin-treated cells. Finally, inhibition of SHP phosphatase activity by NSC87877 abrogated B. pertussis survival inside murine macrophages. These results reveal that an as yet unknown cAMP-activated signaling pathway controls SHP-1 phosphatase activity and may regulate numerous receptor signaling pathways in leukocytes. Hijacking of SHP-1 by CyaA action then enables B. pertussis to evade NO-mediated killing in sentinel cells of innate immunity.
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Pricing and availability is not currently available.
Sigma-Aldrich
Sulfanilamide, puriss. p.a., ≥98% (calc. to the dried substance)
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium chloride solution, 0.85%
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Phosphoric acid solution, 85 wt. % in H2O, FCC, FG
Pricing and availability is not currently available.
Sigma-Aldrich
Phosphoric acid solution, NMR reference standard, 85% in D2O (99.9 atom % D), NMR tube size 4.2 mm × 8 in. , WGS-5BL Coaxial NMR tube
Pricing and availability is not currently available.
Sigma-Aldrich
AIDA, solid
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Pricing and availability is not currently available.
Sigma-Aldrich
3-Isobutyl-1-methylxanthine, ≥99% (HPLC), powder
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium Fluoride Solution
Pricing and availability is not currently available.
Sigma-Aldrich
3-Isobutyl-1-methylxanthine, ≥99%, BioUltra
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium chloride, tablet
Pricing and availability is not currently available.
Sigma-Aldrich
Sulfanilamide, ≥98%
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium chloride solution, 5 M
Pricing and availability is not currently available.
SAFC
Sodium chloride solution, 5 M
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Pricing and availability is not currently available.
Sigma-Aldrich
Phosphoric acid-16O4 solution, 70 wt. % in D2O, 99.9 atom % 16O
Pricing and availability is not currently available.
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
DL-Tyrosine, 99%
Pricing and availability is not currently available.
Sigma-Aldrich
Phosphoric acid, crystalline, ≥99.999% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Phosphoric acid, ≥85 wt. % in H2O, ≥99.999% trace metals basis
Pricing and availability is not currently available.