Merck
Get up to 22% off for Pi Day until 3/26.Save Now
  • miR-221/222 induces pancreatic cancer progression through the regulation of matrix metalloproteinases.
miR-221/222 induces pancreatic cancer progression through the regulation of matrix metalloproteinases.
Oncotarget (2015-04-18)
Qinhong Xu, Pei Li, Xin Chen, Liang Zong, Zhengdong Jiang, Ligang Nan, Jianjun Lei, Wanxing Duan, Dong Zhang, Xuqi Li, Huanchen Sha, Zheng Wu, Qingyong Ma, Zheng Wang
ABSTRACT
MicroRNAs are involved in the initiation and progression of pancreatic cancer. In this study, we showed that miR-221/222 is overexpressed in pancreatic cancer. MiR-221/222 overexpression significantly promoted pancreatic cancer cell proliferation and invasion while inhibiting apoptosis. The expression of the matrix metalloproteinases (MMPs) MMP-2 and MMP-9 was increased in miR-221/222 mimic-transfected pancreatic cancer cells. Validation experiments identified TIMP-2 as a direct target of miR-221/222. These data indicate that overexpressed miR-221/222 may play an oncogenic role in pancreatic cancer by inducing the expression of MMP-2 and MMP-9, thus leading to cancer cell invasion.
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
MISSION® esiRNA, targeting human TIMP2
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Timp2
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Propidium iodide solution
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, Molecular Biology
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, Vetec, reagent grade, 99%
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)