Merck
  • Clinical, biochemical and histological study of the effect of antimicrobial photodynamic therapy on oral mucositis induced by 5-fluorouracil in hamsters.
Clinical, biochemical and histological study of the effect of antimicrobial photodynamic therapy on oral mucositis induced by 5-fluorouracil in hamsters.
Photodiagnosis and photodynamic therapy (2015-01-24)
Érika de Paula da Cruz, Luana Campos, Filipi da Silva Pereira, Gabriela Campos Magliano, Bernar Monteiro Benites, Victor Elias Arana-Chavez, Rafael Yagüe Ballester, Alyne Simões
ABSTRACT
Oral mucositis (OM) is a debilitating side effect of chemotherapy, which can be relieved by phototherapy. Antimicrobial photodynamic therapy (aPDT) may be used for the treatment of OM, when infection is present. However, there are no studies showing that aPDT affects tissue repair process when used in the treatment of lesions caused by OM. This work aims to evaluate the effect of aPDT in healing OM induced by 5-Fluorouracil (5-FU). Two hundred forty-five hamsters were divided into two groups, control (C) and experimental, which were subdivided into 4 subgroups (Ch, ChP, ChL, aPDT). C group received only the vehicle of chemotherapy and anesthesia, whereas all animals of the experimental groups received anesthesia and chemotherapy agent 5-FU to induce OM. Ch group received no OM treatment; ChP group received an application of methylene blue (MB) 0.01%; ChL received irradiation with low-power-laser (LPL-660 nm/120 J /cm(2)/40 mW/4.4 J per point); and aPDT received MB and LPL irradiation. OM Clinical severity were daily assessed by a blinded examiner. The animals were sacrificed after 5, 7 and 10 days of experiment and their oral mucosa were removed for biochemical (enzymatic activity of SOD and catalase) and histological analyzes (light microscopy). After statistical analysis was performed, results showed that aPDT reduced the severity of OM on the tenth day of the experiment, when compared to the initial OM score (p < 0.05), as well as increased keratinization with organized collagen deposition in the lamina propria. In conclusion, aPDT can be safely used in animals with infected OM because it does not affect lesion-repairing processes.
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 70% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 8% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 25% in H2O, specially purified for use as an electron microscopy fixative
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 50% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Glutaraldehyde solution, 50% in H2O, suitable for photographic applications
Sigma-Aldrich
Glutaraldehyde solution, Grade II, 25% in H2O
Sigma-Aldrich
Formaldehyde solution, for molecular biology, 36.5-38% in H2O
Sigma-Aldrich
Formaldehyde solution, meets analytical specification of USP, ≥34.5 wt. %
Sigma-Aldrich
Formaldehyde solution, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Sigma-Aldrich
Formaldehyde solution, for molecular biology, BioReagent, ≥36.0% in H2O (T)
Sigma-Aldrich
Glutaraldehyde solution, 50 wt. % in H2O
Sigma-Aldrich
Glutaric dialdehyde solution, 50 wt. % in H2O, FCC
Sigma-Aldrich
Formaldehyde-12C solution, 20% in H2O, 99.9 atom % 12C
Sigma-Aldrich
Fluorouracil, meets USP testing specifications
Sigma-Aldrich
5-Fluorouracil, ≥99% (HPLC), powder