Merck
  • Establishment and characterization of a fish-cell line from the brain of Japanese flounder Paralichthys olivaceus.
Establishment and characterization of a fish-cell line from the brain of Japanese flounder Paralichthys olivaceus.
Journal of fish biology (2015-05-15)
Y Zheng, L M Peng, F You, Y X Zou, P J Zhang, S L Chen
ABSTRACT
A new brain-cell line derived from Japanese flounder Paralichthys olivaceus (POBC) was established. POBC was subcultured for 67 passages over the course of 420 days. The cultured cells were primarily epithelioid-like. Chromosome analysis revealed the cell line to possess the normal P. olivaceus diploid karyotype of 2n = 48t (telocentric chromosomes). The cells exhibited the astrocyte marker glial fibrillary acidic protein by immunocytochemistry, and significant fluorescent signals were observed when the cells were transfected with green fluorescent protein reporter plasmid. The established POBC would be ideal material for the study of function of fish ependyma, the central neuroendocrine system and endocrine disruptors in the marine environment.
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Acetic acid, for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, purum, ≥99.0% (TLC)
Supelco
Ethanol standards 10% (v/v), 10 % (v/v) in H2O, analytical standard
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, ≥97.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Supelco
Methanol solution, contains 0.10 % (v/v) formic acid, UHPLC, suitable for mass spectrometry (MS), ≥99.5%