Merck
  • A basis for vaccine development: Comparative characterization of Haemophilus influenzae outer membrane vesicles.
A basis for vaccine development: Comparative characterization of Haemophilus influenzae outer membrane vesicles.
International journal of medical microbiology : IJMM (2015-01-17)
Sandro Roier, Thomas Blume, Lisa Klug, Gabriel E Wagner, Wael Elhenawy, Klaus Zangger, Ruth Prassl, Joachim Reidl, Günther Daum, Mario F Feldman, Stefan Schild
ABSTRACT
Outer membrane vesicles (OMVs) are spherical and bilayered particles that are naturally released from the outer membrane (OM) of Gram-negative bacteria. They have been proposed to possess several biological roles in pathogenesis and interbacterial interactions. Additionally, OMVs have been suggested as potential vaccine candidates against infections caused by pathogenic bacteria like Haemophilus influenzae, a human pathogen of the respiratory tract. Unfortunately, there is still a lack of fundamental knowledge regarding OMV biogenesis, protein sorting into OMVs, OMV size and quantity, as well as OMV composition in H. influenzae. Thus, this study comprehensively characterized and compared OMVs and OMs derived from heterologous encapsulated as well as nonencapsulated H. influenzae strains. Semiquantitative immunoblot analysis revealed that certain OM proteins are enriched or excluded in OMVs suggesting the presence of regulated protein sorting mechanisms into OMVs as well as interconnected OMV biogenesis mechanisms in H. influenzae. Nanoparticle tracking analysis, transmission electron microscopy, as well as protein and lipooligosaccharide quantifications demonstrated that heterologous H. influenzae strains differ in their OMV size and quantity. Lipidomic analyses identified palmitic acid as the most abundant fatty acid, while phosphatidylethanolamine was found to be the most dominant phospholipid present in OMVs and the OM of all strains tested. Proteomic analysis confirmed that H. influenzae OMVs contain vaccine candidate proteins as well as important virulence factors. These findings contribute to the understanding of OMV biogenesis as well as biological roles of OMVs and, in addition, may be important for the future development of OMV based vaccines against H. influenzae infections.
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium hydroxide, BioUltra, for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, for molecular biology, 10 M in H2O
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Tris(tert-butoxy)silanol, 99.999%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Methyl hexacosanoate, ≥99% (capillary GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
Supelco
DL-Dithiothreitol solution, 1 M in H2O
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
p-Coumaric acid, ≥98.0% (HPLC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
Sodium hydroxide, ultra dry, powder or crystals, 99.99% trace metals basis
Sigma-Aldrich
Dimethyl sulfoxide, Vetec, reagent grade, 99%
Sigma-Aldrich
Iodoacetamide, Vetec, reagent grade, 98%
SAFC
Iodoacetamide
Sigma-Aldrich
Sodium hydroxide-16O solution, 20 wt. % in H216O, 99.9 atom % 16O
Sigma-Aldrich
Luminol, 97%
Sigma-Aldrich
Sucrose, puriss., meets analytical specification of Ph. Eur., BP, NF
Sigma-Aldrich
Sucrose, BioUltra, for molecular biology, ≥99.5% (HPLC)
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 10% in H2O