Merck
  • Optimization and Performance Assessment of the Chorion-Off [Dechorinated] Zebrafish Developmental Toxicity Assay.
Optimization and Performance Assessment of the Chorion-Off [Dechorinated] Zebrafish Developmental Toxicity Assay.
Toxicological sciences : an official journal of the Society of Toxicology (2015-04-17)
Julieta M Panzica-Kelly, Cindy X Zhang, Karen A Augustine-Rauch
ABSTRACT
The Dechorinated Zebrafish Embryo Developmental toxicity assay was originally developed from a training set of 31 compounds and reported to be 87% concordant with in vivo teratogenicity data (Brannen, K. C., Panzica-Kelly, J. M., Danberry, T. L., and Augustine-Rauch, K. A. (2010). Development of a zebrafish embryo teratogenicity assay and quantitative prediction model. Birth Defects Res. 89, 66-77.). The assay includes scoring larva treated in a concentration range for malformations of specific morphological structures/organ systems. The model includes identifying a no-adverse-effect-level (NOAEL) and the concentration resulting in 25% lethality (LC25) at 5 days postfertilization. An LC25/NOAEL ratio ≥10 classifies a compound positive for teratogenic potential. A consortium effort evaluated a modified version of this assay which involved enzymatic chorion treatment instead of manual dissection and used experimental replicates for final classification. The modified assay achieved an 85% overall predictivity (Gustafson, A. L., Stedman, D. B., Ball, J., Hillegass, J. M., Flood, A., Zhang, C. X., Panzica-Kelly, J., Cao, J., Coburn, A., Enright, B. P., et al. (2012). Inter-laboratory assessment of a harmonized zebrafish developmental toxicology assay - progress report on phase I. Reprod. Toxicol. 33, 155-164.). The objective of this study was to perform a thorough performance evaluation of the dechorinated assay by repeating the original training set and testing additional compounds in experimental replicates. When the initial training set was repeated with inclusion of experimental replicates, the overall predictivity was 83%. Model performance was tested with an additional 34 compounds and achieved overall predictivity of 74%. When the training and test sets were combined (63 compounds) the assay's final sensitivity was 83% and the specificity was 71%. Total predictivity was 78% with relatively balanced predictivity for nonteratogens (77%) and teratogens (78%). The chorion-off assay achieved superior sensitivity (83%) compared with sensitivity (63-74%) reported by consortium efforts testing a similar assay with chorion-intact embryos (Ball, J. S., Stedman, D. B., Hillegass, J. M., Zhang, C. X., Panzica-Kelly, J., Coburn, A., Enright, B. P., Tornesi, B., Amouzadeh, H. R., Hetheridge, M., et al. (2014). Fishing for teratogens: a consortium effort for a harmonized zebrafish developmental toxicology assay. Toxicol. Sci. 139, 210-219.). Additional protocol modifications were made to increase assay throughput.
Sigma-Aldrich
Dimethyl sulfoxide, Vetec, reagent grade, 99%
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture