Merck
  • Pancreatic cancer exosomes initiate pre-metastatic niche formation in the liver.
Pancreatic cancer exosomes initiate pre-metastatic niche formation in the liver.
Nature cell biology (2015-05-20)
Bruno Costa-Silva, Nicole M Aiello, Allyson J Ocean, Swarnima Singh, Haiying Zhang, Basant Kumar Thakur, Annette Becker, Ayuko Hoshino, Milica Tešić Mark, Henrik Molina, Jenny Xiang, Tuo Zhang, Till-Martin Theilen, Guillermo García-Santos, Caitlin Williams, Yonathan Ararso, Yujie Huang, Gonçalo Rodrigues, Tang-Long Shen, Knut Jørgen Labori, Inger Marie Bowitz Lothe, Elin H Kure, Jonathan Hernandez, Alexandre Doussot, Saya H Ebbesen, Paul M Grandgenett, Michael A Hollingsworth, Maneesh Jain, Kavita Mallya, Surinder K Batra, William R Jarnagin, Robert E Schwartz, Irina Matei, Héctor Peinado, Ben Z Stanger, Jacqueline Bromberg, David Lyden
ABSTRACT
Pancreatic ductal adenocarcinomas (PDACs) are highly metastatic with poor prognosis, mainly due to delayed detection. We hypothesized that intercellular communication is critical for metastatic progression. Here, we show that PDAC-derived exosomes induce liver pre-metastatic niche formation in naive mice and consequently increase liver metastatic burden. Uptake of PDAC-derived exosomes by Kupffer cells caused transforming growth factor β secretion and upregulation of fibronectin production by hepatic stellate cells. This fibrotic microenvironment enhanced recruitment of bone marrow-derived macrophages. We found that macrophage migration inhibitory factor (MIF) was highly expressed in PDAC-derived exosomes, and its blockade prevented liver pre-metastatic niche formation and metastasis. Compared with patients whose pancreatic tumours did not progress, MIF was markedly higher in exosomes from stage I PDAC patients who later developed liver metastasis. These findings suggest that exosomal MIF primes the liver for metastasis and may be a prognostic marker for the development of PDAC liver metastasis.
Sigma-Aldrich
MISSION® esiRNA, targeting human MIF
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
Tamoxifen, ≥99%
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Supelco
Methanol solution, contains 0.10 % (v/v) formic acid, UHPLC, suitable for mass spectrometry (MS), ≥99.5%
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Mif