Merck
  • Impact of local endothelial shear stress on neointima and plaque following stent implantation in patients with ST-elevation myocardial infarction: A subgroup-analysis of the COMFORTABLE AMI-IBIS 4 trial.
Impact of local endothelial shear stress on neointima and plaque following stent implantation in patients with ST-elevation myocardial infarction: A subgroup-analysis of the COMFORTABLE AMI-IBIS 4 trial.
International journal of cardiology (2015-04-02)
Christos V Bourantas, Lorenz Räber, Serge Zaugg, Antonis Sakellarios, Masanori Taniwaki, Dik Heg, Aris Moschovitis, Maria Radu, Michail I Papafaklis, Fanis Kalatzis, Katerina K Naka, Dimitrios I Fotiadis, Lampros K Michalis, Patrick W Serruys, Hector M Garcia Garcia, Stephan Windecker
ABSTRACT
Numerous studies have demonstrated an association between endothelial shear stress (ESS) and neointimal formation after stent implantation. However, the role of ESS on the composition of neointima and underlying plaque remains unclear. Patients recruited in the Comfortable AMI-IBIS 4 study implanted with bare metal stents (BMS) or biolimus eluting stents (BES) that had biplane coronary angiography at 13 month follow-up were included in the analysis. The intravascular ultrasound virtual-histology (IVUS-VH) and the angiographic data were used to reconstruct the luminal surface, and the stent in the stented segments. Blood flow simulation was performed in the stent surface, which was assumed to represent the luminal surface at baseline, to assess the association between ESS and neointima thickness. The predominant ESS was estimated in 3-mm segments and was correlated with the amount of neointima, neointimal tissue composition, and with the changes in the underlying plaque burden and composition. Forty three patients (18 implanted with BMS and 25 with BES) were studied. In both stent groups negative correlations were noted between ESS and neointima thickness in BMS (P < 0.001) and BES (P = 0.002). In BMS there was a negative correlation between predominant ESS and the percentage of the neointimal necrotic core component (P = 0.015). In BES group, the limited neointima formation did not allow evaluation of the effect of ESS on its tissue characteristics. ESS did not affect vessel wall remodeling and the plaque burden and composition behind BMS (P > 0.10) and BES (P > 0.45). ESS determines neointimal formation in both BMS and BES and affects the composition of the neointima in BMS. Conversely, ESS does not impact the plaque behind struts irrespective of stent type throughout 13 months of follow-up.
Sigma-Aldrich
BES, Vetec, reagent grade, ≥99%
Sigma-Aldrich
BES, ≥99.0% (titration)