Merck
  • Bioconversion of xylose, hexoses and biomass to ethanol by a new isolate of the white rot basidiomycete Trametes versicolor.
Bioconversion of xylose, hexoses and biomass to ethanol by a new isolate of the white rot basidiomycete Trametes versicolor.
SpringerPlus (2014-03-14)
Kenji Okamoto, Atsushi Uchii, Ryuichi Kanawaku, Hideshi Yanase
ABSTRACT
Second-generation bioethanol production requires the development of economically feasible and sustainable processes that use renewable lignocellulosic biomass as a starting material. However, the microbial fermentation of xylose, which is the principal pentose sugar in hemicellulose, is a limiting factor in developing such processes. Here, a strain of the white rot basidiomycete Trametes versicolor that was capable of efficiently fermenting xylose was newly isolated and characterized. This strain, designated KT9427, was capable of assimilating and converting xylose to ethanol under anaerobic conditions with a yield of 0.44 g ethanol per 1 g of sugar consumed. In culture medium containing low yeast extract concentrations, xylose consumption and ethanol productivity were enhanced. Adjusting the initial pH between 3.0 and 5.0 did not markedly influence xylose fermentation. T. versicolor KT9427 also produced ethanol from glucose, mannose, fructose, cellobiose and maltose at yields ranging from 0.45 to 0.49 g ethanol per 1 g of sugar consumed. In addition, strain KT9427 exhibited favourable conversion of non-pretreated starch, cellulose, xylan, wheat bran and rice straw into ethanol compared to common recombinant yeast strains. Taken together, the present findings suggest that T. versicolor KT9427 is a promising candidate for environmentally friendly ethanol production directly from lignocellulosic biomass.
Sigma-Aldrich
D-Glucose-12C6, 16O6, 99.9 atom % 16O, 99.9 atom % 12C
Sigma-Aldrich
D-(+)-Mannose, BioUltra, ≥99.5% (sum of enantiomers, HPLC)
Sigma-Aldrich
D-(+)-Glucose, BioUltra, anhydrous, ≥99.5% (sum of enantiomers, HPLC)
Sigma-Aldrich
D-(+)-Mannose, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
D-(−)-Fructose, ≥99% (HPLC), BioXtra
Sigma-Aldrich
D-(−)-Fructose, 98.0-102.0% dry basis, meets USP testing specifications
Sigma-Aldrich
D-(−)-Fructose, ≥99% (HPLC)
Sigma-Aldrich
D-(−)-Fructose, ≥99% (HPLC), BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
D-(+)-Glucose, ACS reagent
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, suitable for mouse embryo cell culture, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.5%
Sigma-Aldrich
D-(+)-Glucose, Hybri-Max, powder, BioReagent, suitable for hybridoma
Sigma-Aldrich
D-(+)-Mannose, ≥99% (GC), wood
Sigma-Aldrich
D-(−)-Fructose, BioUltra, ≥99.0% (HPLC)
Sigma-Aldrich
D-(+)-Mannose, synthetic, ≥99% (GC)
Sigma-Aldrich
D-(+)-Glucose, Vetec, reagent grade, ≥99.5% (HPLC)
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
D-(+)-Xylose, ≥99%
Sigma-Aldrich
Magnesium sulfate solution, BioUltra, for molecular biology
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Potassium phosphate monobasic, BioUltra, for molecular biology, anhydrous, ≥99.5% (T)
Sigma-Aldrich
D-(+)-Xylose, BioUltra, ≥99.0% (sum of enantiomers, HPLC)
Sigma-Aldrich
Magnesium sulfate solution, for molecular biology, 1.00 M±0.04 M
Supelco
Ethanol standards 10% (v/v), 10 % (v/v) in H2O, analytical standard
Sigma-Aldrich
Dextrose, 97.5-102.0% anhydrous basis, meets EP, BP, JP, USP testing specifications
Sigma-Aldrich
Potassium phosphate monobasic, for molecular biology, ≥98.0%
Sigma-Aldrich
Potassium phosphate monobasic, powder, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0%
Sigma-Aldrich
Potassium phosphate monobasic, ReagentPlus®
Sigma-Aldrich
D-(+)-Xylose, BioXtra, ≥99% (GC)