Merck
  • Co-Doping of Activated Graphene for Synergistically Enhanced Electrocatalytic Oxygen Reduction Reaction.
Co-Doping of Activated Graphene for Synergistically Enhanced Electrocatalytic Oxygen Reduction Reaction.
ChemSusChem (2015-11-14)
Mahbub Hassan, Enamul Haque, Andrew I Minett, Vincent G Gomes
ABSTRACT
Doping of graphene has emerged as a key strategy to improve the electrocatalytic performance of the oxygen reduction reaction (ORR). Activated graphene co-doped with iodine and nitrogen atoms (NIG) was developed in this work using a facile scalable approach. The onset potential, current density, and four-electron reduction pathway of the newly developed catalyst were significantly improved. The charge-transfer resistance of co-doped NIG was found to be much lower than nitrogen-doped graphene (NG); furthermore, the stability of NIG and its resistance to methanol crossover were also improved. The synergistically enhanced ORR performance of NIG was found to be a result of a high strain and size advantage of the larger iodine atom clusters (compared to nitrogen), which facilitate the simultaneous enrichment of anode electrons and O2 and H2 O molecule transport at catalytic sites, inducing four-electron transfer in a single step. These results are promising for application in alkaline fuel cells.
Sigma-Aldrich
Water, ACS reagent
Sigma-Aldrich
Water, Deionized
Sigma-Aldrich
Water, PCR Reagent
Sigma-Aldrich
Water, for embryo transfer, sterile-filtered, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Water, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Water, Nuclease-Free Water, for Molecular Biology
Sigma-Aldrich
Water, for molecular biology, sterile filtered
Sigma-Aldrich
Water, for cell biology, sterile ultrafiltered
Sigma-Aldrich
Water, BioPerformance Certified
Sigma-Aldrich
E-Toxate Water, endotoxin, free
Sigma-Aldrich
Iodine, puriss., ≥99.5% (RT), particles (round)
Supelco
Iodine, ReagentPlus®, ≥99.8% (titration)
Sigma-Aldrich
Iodine, ReagentPlus®, 99.7% trace metals basis, beads, 1-3 mm
Sigma-Aldrich
Iodine, ACS reagent, ≥99.8%, solid
Sigma-Aldrich
Iodine, flakes, ReagentPlus®, ≥99%
Sigma-Aldrich
Iodine, ≥99.99% trace metals basis
Sigma-Aldrich
Iodine, anhydrous, beads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Iodine, 99.999% trace metals basis
Sigma-Aldrich
Iodine, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.8-100.5%