Merck
  • Redistribution of subcellular calcium and its effect on apoptosis in primary cultures of rat proximal tubular cells exposed to lead.
Redistribution of subcellular calcium and its effect on apoptosis in primary cultures of rat proximal tubular cells exposed to lead.
Toxicology (2015-04-30)
Hong Wang, Zhong-Kun Wang, Peng Jiao, Xu-Ping Zhou, Du-Bao Yang, Zhen-Yong Wang, Lin Wang
ABSTRACT
Previous studies have shown that cytosolic Ca(2+) ([Ca(2+)]c) overload was involved in Pb-induced apoptosis in primary cultures of rat proximal tubular (rPT) cells, but the source of elevated Ca(2+) and the effect of potential subcellular Ca(2+) redistribution on apoptosis are still unknown. In this study, variations of [Ca(2+)]c in two culture media (Ca(2+)-containing and Ca(2+)- free) were analyzed, indicating that Pb-induced elevation of [Ca(2+)]c was primarily generated intracellularly. Fluo-4-AM, dihydro-Rhod-2-AM and Mag-Fluo-4-AM was loaded to Pb-exposed rPT cells to monitor the imaging of Ca(2+) concentrations in the cytoplasm ([Ca(2+)]c), mitochondria ([Ca(2+)]mit) and endoplasmic reticulum (ER) ([Ca(2+)]ER), respectively, under the confocal microscope. Data indicate that elevations of [Ca(2+)]c and [Ca(2+)]mit with depletion of [Ca(2+)]ER were revealed in Pb-treated rPT cells, but this subcellular Ca(2+) redistribution could be significantly suppressed by 2-APB, a specific inhibitor of inositol 1,4,5-trisphosphate receptor (IP3R) that functions to release Ca(2+) from ER stores. Simultaneously, Pb-mediated mitochondrial Ca(2+) overload can be partially suppressed by the cytosolic Ca(2+) chelator BAPTA-AM, suggesting that Ca(2+) uptake into mitochondria occurs via diverse pathways and ER Ca(2+) storage was the chief source. Furthermore, Pb-induced apoptosis was markedly inhibited by 2-APB and BAPTA-AM, respectively. Additionally, elevated IP3 levels with up-regulated IP3R-1 and IP3R-2 (mRNA and protein) levels were revealed in Pb-exposed rPT cells. In summary, IP3R-mediated ER Ca(2+) release promoted the elevations of [Ca(2+)]c and [Ca(2+)]mit in Pb-exposed rPT cells, which played a chief role in apoptosis induced by impaired calcium homeostasis.
Sigma-Aldrich
Sodium borohydride solution, 2.0 M in triethylene glycol dimethyl ether
Sigma-Aldrich
VenPure® SF, powder
Sigma-Aldrich
Sodium borohydride solution, ~12 wt. % in 14 M NaOH
Sigma-Aldrich
N,N,N′,N′-Tetrakis(2-pyridylmethyl)ethylenediamine
Sigma-Aldrich
Sodium borohydride, granular, 10-40 mesh, 98%
Sigma-Aldrich
Sodium borohydride, powder, ≥98.0%
Sigma-Aldrich
Ethylenediamine, BioXtra
Sigma-Aldrich
Ethylenediamine, meets USP testing specifications
Sigma-Aldrich
2-Aminoethyl diphenylborinate, 97%
Sigma-Aldrich
Ethylenediamine, purified by redistillation, ≥99.5%
Sigma-Aldrich
Sodium borohydride, ReagentPlus®, 99%
Sigma-Aldrich
Sodium borohydride, purum p.a., ≥96% (gas-volumetric)
Sigma-Aldrich
Ethylenediamine solution, technical, 75-80%
Sigma-Aldrich
Sodium borohydride, caplets (18 × 10 × 8 mm), 98%
Sigma-Aldrich
Ethylenediamine, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium borohydride, granular, 99.99% trace metals basis