MilliporeSigma
  • A simple method for the determination of organochlorine pollutants and the enantiomers in oil seeds based on matrix solid-phase dispersion.
A simple method for the determination of organochlorine pollutants and the enantiomers in oil seeds based on matrix solid-phase dispersion.
Food chemistry (2015-10-17)
Jing Zhan, Jindong Li, Donghui Liu, Chang Liu, Genggeng Yang, Zhiqiang Zhou, Peng Wang
ABSTRACT
A simple, rapid and effective method was developed based on matrix solid-phase dispersion (MSPD) for the determination of organochlorine pollutants including sixteen organochlorine pesticides (OCPs) and seven polychlorinated biphenyls (PCBs) in oil seeds (peanuts and soybeans). Among the organochlorine pollutants selected, α-HCH, heptachlor, o,p'-DDT, o,p'-DDD, trans-chlordane and cis-chlordane were chiral and their enantiomers were determined by GC-ECD with a chiral column. The MSPD procedure was optimized focusing on the type and amount of dispersion sorbent, co-column sorbent and eluting solvent. Under the optimized condition, good recoveries were obtained in the range of 68.9-103.3% with relative standard deviations (RSD) values below 16.1% in all cases. LODs and LOQs were in range of 0.1-2.0ngg(-1) and 0.4-6.7ngg(-1) respectively. In summary, the method set up was qualified to be used for the determination of the pollutants and the enantiomers in oil seeds.
Sigma-Aldrich
Aluminum oxide, nanopowder, 13 nm primary particle size (TEM), 99.8% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Aluminum oxide, nanopowder, <50 nm particle size (TEM)
Pricing and availability is not currently available.
Sigma-Aldrich
Hexane, HPLC Plus, for HPLC, GC, and residue analysis, ≥95%
Pricing and availability is not currently available.
Sigma-Aldrich
Aluminum oxide, nanoparticles, <50 nm particle size (DLS), 20 wt. % in isopropanol
Pricing and availability is not currently available.
Supelco
Dichloromethane solution, contains 10 % (v/v) methanol
Pricing and availability is not currently available.
Sigma-Aldrich
Dichloromethane, suitable for HPLC, ≥99.9%, contains 40-150 ppm amylene as stabilizer
Pricing and availability is not currently available.
Sigma-Aldrich
2-Methylheptane, 98%
Pricing and availability is not currently available.
Sigma-Aldrich
Dichloromethane, anhydrous, ≥99.8%, contains 40-150 ppm amylene as stabilizer
Pricing and availability is not currently available.
Sigma-Aldrich
Aluminum oxide, 99.997% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Hexane, puriss. p.a., ACS reagent, ≥99.0% (GC)
Pricing and availability is not currently available.
Sigma-Aldrich
Hexane, anhydrous, 95%
Pricing and availability is not currently available.
Sigma-Aldrich
Aluminum oxide, nanoparticles, 30-60 nm particle size (TEM), 20 wt. % in H2O
Pricing and availability is not currently available.
Sigma-Aldrich
Aluminum oxide, single crystal substrate, <0001>
Pricing and availability is not currently available.
Sigma-Aldrich
Aluminum oxide, nanowires, diam. × L 2-6 nm × 200-400 nm
Pricing and availability is not currently available.
Sigma-Aldrich
Dichloromethane, puriss. p.a., ACS reagent, reag. ISO, ≥99.9% (GC)
Pricing and availability is not currently available.
Sigma-Aldrich
Dichloromethane, ACS reagent, ≥99.5%, contains 40-150 ppm amylene as stabilizer
Pricing and availability is not currently available.
Sigma-Aldrich
Hexane, ReagentPlus®, ≥99%
Pricing and availability is not currently available.
Sigma-Aldrich
Aluminum oxide, activated, basic, Brockmann I
Pricing and availability is not currently available.
Sigma-Aldrich
Aluminum oxide, activated, acidic, Brockmann I
Pricing and availability is not currently available.
Sigma-Aldrich
Aluminum oxide, activated, neutral, Brockmann I
Pricing and availability is not currently available.
Sigma-Aldrich
Aluminum oxide, powder, primarily α phase, ≤10 μm avg. part. size, 99.5% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Dichloromethane, contains 40-150 ppm amylene as stabilizer, ACS reagent, ≥99.5%
Pricing and availability is not currently available.
Sigma-Aldrich
Aluminum oxide, Type WN-6, Neutral, Activity Grade Super I
Pricing and availability is not currently available.
Sigma-Aldrich
Dichloromethane, ACS reagent, ≥99.5%, contains 40-150 ppm amylene as stabilizer
Pricing and availability is not currently available.
Sigma-Aldrich
Aluminum oxide, activated, neutral, Brockmann I, free-flowing, Redi-Dri
Pricing and availability is not currently available.
Sigma-Aldrich
Aluminum oxide, activated, acidic, Brockmann I, free-flowing, Redi-Dri
Pricing and availability is not currently available.
Sigma-Aldrich
Dichloromethane, puriss., meets analytical specification of Ph. Eur., NF, ≥99% (GC)
Pricing and availability is not currently available.
Sigma-Aldrich
Aluminum oxide, fused, powder, primarily α-phase, -325 mesh
Pricing and availability is not currently available.
Sigma-Aldrich
Aluminum oxide, powder, 99.99% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Aluminum oxide, fused, powder, primarily α-phase, 100-200 mesh
Pricing and availability is not currently available.