Merck
  • Lipidomic analysis of plasma lipoprotein fractions in myocardial infarction-prone rabbits.
Lipidomic analysis of plasma lipoprotein fractions in myocardial infarction-prone rabbits.
Journal of bioscience and bioengineering (2015-07-15)
Hiroaki Takeda, Tomonari Koike, Yoshihiro Izumi, Takayuki Yamada, Masaru Yoshida, Masashi Shiomi, Eiichiro Fukusaki, Takeshi Bamba
ABSTRACT
Lipids play important roles in the body and are transported to various tissues via lipoproteins. It is commonly assumed that alteration of lipid levels in lipoproteins leads to dyslipidemia and serious diseases such as coronary artery disease (CAD). However, lipid compositions in each lipoprotein fraction induced by lipoprotein metabolism are poorly understood. Lipidomics, which involves the comprehensive and quantitative analysis of lipids, is expected to provide valuable information regarding the pathogenic mechanism of CAD. Here, we performed a lipidomic analysis of plasma and its lipoprotein fractions in myocardial infarction-prone Watanabe heritable hyperlipidemic (WHHLMI) rabbits. In total, 172 lipids in plasma obtained from normal and WHHLMI rabbits were quantified with high throughput and accuracy using supercritical fluid chromatography hybrid quadrupole-Orbitrap mass spectrometry (SFC/Q-Orbitrap-MS). Plasma levels of each lipid class (i.e., phosphatidylcholine, phosphatidylethanolamine, phosphatidylinositol, lysophosphatidylcholine, lysophosphatidylethanolamine, sphingomyelin, ceramide, triacylglycerol, diacylglycerol, and cholesterol ester, except for free fatty acids) in 21-month-old WHHLMI rabbits were significantly higher than those in normal rabbits. High levels of functional lipids, such as alkyl-phosphatidylcholines, phospholipids including ω-6 fatty acids, and plasmalogens, were also observed in WHHLMI rabbit plasma. In addition, high-resolution lipidomic analysis using very low density lipoprotein (VLDL) and low density lipoprotein (LDL) provided information on the specific molecular species of lipids in each lipoprotein fraction. In particular, higher levels of phosphatidylethanolamine plasmalogens were detected in LDL than in VLDL. Our lipidomics approach for plasma lipoprotein fractions will be useful for in-depth studies on the pathogenesis of CAD.
Product Description
SAFC
Cholesterol, from sheep wool, Controlled origin, meets USP/NF testing specifications
Sigma-Aldrich
Sodium hydroxide, BioXtra, ≥98% (acidimetric), pellets (anhydrous)
Sigma-Aldrich
Sodium hydroxide, puriss., meets analytical specification of Ph. Eur., BP, NF, E524, 98-100.5%, pellets
Sigma-Aldrich
Sodium hydroxide, ultra dry, powder or crystals, 99.99% trace metals basis
Sigma-Aldrich
Ammonium acetate, Vetec, reagent grade, 97%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Sodium hydroxide solution, 5.0 M
Sigma-Aldrich
Sodium hydroxide, puriss. p.a., ACS reagent, K ≤0.02%, ≥98.0% (T), pellets
Sigma-Aldrich
Sodium bromide, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%
Sigma-Aldrich
Chloroform, ReagentPlus®, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
SyntheChol® NS0 Supplement, 500 ×, synthetic cholesterol, animal component-free, aqueous solution, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Sodium bromide, BioXtra, ≥99.0%
Sigma-Aldrich
Cholesterol, Sigma Grade, ≥99%
Sigma-Aldrich
Ammonium acetate, reagent grade, ≥98%
Supelco
Chloroform, suitable for HPLC, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, meets analytical specification of Ph. Eur., BP, USP, FCC, 99.0-101.0%
Sigma-Aldrich
Sodium hydroxide, BioUltra, for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Sodium bromide, BioUltra, ≥99.0% (AT)
Sigma-Aldrich
Sodium bromide, ≥99.99% trace metals basis
Sigma-Aldrich
Chloroform, ACS reagent, ≥99.8%, contains amylenes as stabilizer
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Ammonium acetate, 99.999% trace metals basis
Sigma-Aldrich
Chloroform, ACS reagent, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer