Merck
  • Homocysteine metabolism, growth performance, and immune responses in suckling and weanling piglets.
Homocysteine metabolism, growth performance, and immune responses in suckling and weanling piglets.
Journal of animal science (2014-11-22)
I Audet, C L Girard, M Lessard, L Lo Verso, F Beaudoin, J J Matte
ABSTRACT
Homocysteine (Hcy), an intermediary sulfur AA, is recognized as a powerful prooxidant with deleterious effects on physiological and immune functions. In piglets, there is an acute 10-fold increase of plasma concentrations of homocysteine (pHcy) during the first 2 wk of life. This project aimed to maximize pHcy variations within physiological ranges using typical supplies of folates and vitamin B12 (B12) to sows and piglets. Growth, immune response, and Hcy metabolism of piglets were studied until piglets reached 56 d of age. Third-parity sows were randomly assigned to a 2 × 2 split-plot design with 2 dietary treatments during gestation and lactation, S(-) (1 mg/kg folates and 20 µg/kg B12, n = 15) and S(+) (10-fold S(-) levels, n = 16), and 2 treatments to piglets within each half litter, intramuscular injections (150 µg) of B12 (P(+)) at d 1 and 21 (weaning) and saline (P(-)). Within each litter of 12 piglets, 3 P(+) and 3 P(-) piglets were studied for growth and Hcy metabolism, and the others were studied for immune responses. During lactation, plasma B12 decreased and was transiently greater in S(+) vs. S(-) piglets on d 1 and P(+) vs. P(-) piglets on d 7 (sow treatment × age and piglet treatment × age; P < 0.05). From 14 to 21 d of age, pHcy was 33% lower in S(+)P(+) vs. S(-)P(-) piglets (sow treatment × piglet treatment interaction; P < 0.05). At 56 d of age, hepatic B12 was greater and pHcy was lower for P(+) vs. P(-) piglets (P < 0.05). No treatment effect was observed on growth except for a lower postweaning G:F in S(+)P(-) piglets than in others (sow treatment × piglet treatment interaction; P < 0.05). Positive correlations were observed between pHcy and growth (r > 0.29, P < 0.05) before and after weaning. Antibody responses to ovalbumin and serum tumor necrosis factor-α were not affected by treatments, but postweaning serum IL-8 peaked earlier in S(-)P(-) vs. S(+)P(+) piglets (piglet treatment × age; sow treatment × piglet treatment interaction, P < 0.05). Proliferation of lymphocytes in response to the mitogen concanavalin A tended to be lower in culture media supplemented with sera from S(-) vs. S(+) piglets (P = 0.081) and P(-) vs. P(+) piglets (P = 0.098), and the reduction of response was more marked (P < 0.05) with high (>21 µM) compared to medium (17 to 21 µM) or low (<17 µM) pHcy. In conclusion, the present vitamin supplements to sows and/or piglets produced variations of pHcy that were not apparently harmful for growth performance of piglets. The greater pHcy, particularly prevalent in S(-) and/or P(-) piglets, had negative effects on some indicators of immune responses, suggesting that these young animals may be immunologically more fragile.
Sigma-Aldrich
Formaldehyde-12C solution, 20% in H2O, 99.9 atom % 12C
Sigma-Aldrich
Vitamin B12, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
Vitamin B12, ≥98%
Sigma-Aldrich
Formaldehyde solution, for molecular biology, 36.5-38% in H2O
Sigma-Aldrich
5(6)-Carboxyfluorescein diacetate N-succinimidyl ester, BioReagent, suitable for fluorescence, ≥90% (HPLC)
Sigma-Aldrich
5-Carboxy-fluorescein diacetate N-succinimidyl ester, for fluorescence, ≥95.0% (HPLC)
Sigma-Aldrich
Formaldehyde solution, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Sigma-Aldrich
Formaldehyde solution, meets analytical specification of USP, ≥34.5 wt. %
Sigma-Aldrich
Formaldehyde solution, for molecular biology, BioReagent, ≥36.0% in H2O (T)
Sigma-Aldrich
Vitamin B12, Vetec, reagent grade, ≥98%
Sigma-Aldrich
Cyanocobalamin, meets USP testing specifications