Merck
  • Pharmacological evaluation of a novel phosphodiesterase 10A inhibitor in models of antipsychotic activity and cognition.
Pharmacological evaluation of a novel phosphodiesterase 10A inhibitor in models of antipsychotic activity and cognition.
Pharmacology, biochemistry, and behavior (2015-05-20)
Philip G Jones, Michael C Hewitt, John E Campbell, Maria S Quinton, Sharon Engel, Robert Lew, Una Campbell, Douglas F Burdi
ABSTRACT
In this study, we report the pharmacological effects of a novel PDE10A inhibitor, SEP-39. SEP-39 is a potent (1.0nM) inhibitor of human PDE10A in vitro, with good selectivity (>16000-fold) against other PDEs. In an in vivo occupancy study, the RO50 value was determined to be 0.7mg/kg (p.o.), corresponding to plasma and brain exposures of 28ng/mL and 43ng/g, respectively. Using microdialysis, we show that 3mg/kg (p.o.) SEP-39 significantly increased rat striatal cGMP concentrations. Furthermore, SEP-39 inhibits PCP-induced hyperlocomotion at doses of 1 and 3mg/kg (p.o.) corresponding to 59-86% occupancy. At similar doses in a catalepsy study, the time on the bar was increased but the maximal effect was less than that seen with haloperidol. In an EEG study, 3 and 10mg/kg (p.o.) SEP-39 suppressed REM intensity and increased the latency to REM sleep. We also demonstrate the procognitive effects of SEP-39 in the rat novel object recognition assay. These effects appear to require less PDE10A inhibition than the reversal of PCP-induced hyperlocomotion or EEG effects, as improvements in recognition index were seen at doses of 0.3mg/kg and above. Our data demonstrate that SEP-39 is a potent, orally active PDE10A inhibitor with therapeutic potential in a number of psychiatric indications.
Sigma-Aldrich
Risperidone, ≥98% (HPLC), powder
Sigma-Aldrich
Adenosine 3′,5′-cyclic monophosphate, ≥98.5% (HPLC), powder
Sigma-Aldrich
Adenosine 3′,5′-cyclic monophosphate tris salt, ≥97% (HPLC), powder
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Dimethyl sulfoxide, Vetec, reagent grade, 99%