Merck
  • The hypoxia-related microRNA miR-199a-3p displays tumor suppressor functions in ovarian carcinoma.
The hypoxia-related microRNA miR-199a-3p displays tumor suppressor functions in ovarian carcinoma.
Oncotarget (2015-04-04)
Yasuto Kinose, Kenjiro Sawada, Koji Nakamura, Ikuko Sawada, Aska Toda, Erika Nakatsuka, Kae Hashimoto, Seiji Mabuchi, Kazuhiro Takahashi, Hirohisa Kurachi, Ernst Lengyel, Tadashi Kimura
ABSTRACT
During the dissemination of ovarian cancer cells, the cells float in the peritoneal cavity without access to a vascular supply and so are exposed to hypoxic conditions, which may cause the ovarian cancer cells to acquire a more aggressive and malignant phenotype. In this study, we screened microRNAs (miRNAs) to identify those that displayed altered expression patterns under hypoxic conditions and then analyzed their functional roles in ovarian cancer progression. miRNA PCR arrays performed on cells from 2 ovarian cancer cell lines (CaOV3 and RMUG-S) revealed miR-199a-3p as one of the miRNAs that are downregulated under hypoxia. In silico analyses indicated that MET is one of the target genes for miR-199a-3p; subsequently, miR-199a-3p expression was found to be inversely correlated with c-Met expression in ovarian cancer. Transfection of precursor miR-199a-3p into ovarian cancer cells reduced c-Met expression and inhibited the phosphorylation of c-Met, extracellular signal-regulated kinase, and AKT; in addition, proliferation, adhesion, and invasiveness were inhibited. Moreover, overexpression of miR-199a-3p in cancer cells significantly suppressed peritoneal dissemination in a xenograft model. In summary, the hypoxia-related microRNA miR-199a-3p drastically inhibits ovarian cancer progression through the downregulation of c-Met expression. Therefore, miR-199a-3p is a potential target for treating ovarian cancer dissemination.
Sigma-Aldrich
Potassium hydride, in paraffin
Sigma-Aldrich
Potassium, chunks (in mineral oil), 98% trace metals basis
Sigma-Aldrich
o-Xylene, anhydrous, 97%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Potassium hydride, 30 wt % dispersion in mineral oil
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Supelco
Ethanol standards 10% (v/v), 10 % (v/v) in H2O, analytical standard