Merck
  • Sequential Upregulation of Superoxide Dismutase 2 and Heme Oxygenase 1 by tert-Butylhydroquinone Protects Mitochondria during Oxidative Stress.
Sequential Upregulation of Superoxide Dismutase 2 and Heme Oxygenase 1 by tert-Butylhydroquinone Protects Mitochondria during Oxidative Stress.
Molecular pharmacology (2015-06-18)
Jiahong Sun, Xuefang Ren, James W Simpkins
ABSTRACT
Oxidative stress is linked to mitochondrial dysfunction in aging and neurodegenerative conditions. The transcription factor nuclear factor E2-related factor 2 (Nrf2)-antioxidant response element (ARE) regulates intracellular antioxidative capacity to combat oxidative stress. We examined the effect of tert-butylhydroquinone (tBHQ), an Nrf2-ARE signaling pathway inducer, on mitochondrial function during oxidative challenge in neurons. tBHQ prevented glutamate-induced cytotoxicity in an HT-22 neuronal cell line even with an 8-hour exposure delay. tBHQ blocked glutamate-induced intracellular reactive oxygen species (ROS) and mitochondrial superoxide accumulation. It also protected mitochondrial function under glutamate toxicity, including maintaining mitochondrial membrane potential, mitochondrial Ca(2+) hemostasis, and mitochondrial respiration. Glutamate-activated, mitochondria-mediated apoptosis was inhibited by tBHQ as well. In rat primary cortical neurons, tBHQ protected cells from both glutamate and buthionine sulfoximine toxicity. We found that tBHQ scavenged ROS and induced a rapid upregulation of superoxide dismutase 2 (SOD2) expression and a delayed upregulation of heme oxygenase 1 (HO-1) expression. In HT-22 cells with a knockdown of SOD2 expression, delayed treatment with tBHQ failed to prevent glutamate-induced cell death. Briefly, tBHQ rescues mitochondrial function by sequentially increasing SOD2 and HO-1 expression during glutamate-mediated oxidative stress. This study is the first to demonstrate the role of tBHQ in preserving mitochondrial function during oxidative challenge and provides a clinically relevant argument for using tBHQ against acute neuron-compromising conditions.
Sigma-Aldrich
HEPES buffer solution, 1 M in H2O
Sigma-Aldrich
Calcein AM solution, 4 mM in DMSO, ≥90% (HPLC), solution
Sigma-Aldrich
MISSION® esiRNA, targeting human SOD2
Sigma-Aldrich
Tetramethylrhodamine ethyl ester perchlorate, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
L-(−)-Glucose, ≥99%
Sigma-Aldrich
L-Glutathione reduced, BioXtra, ≥98.0%
Sigma-Aldrich
L-Glutathione reduced, suitable for cell culture, BioReagent, ≥98.0%, powder
Sigma-Aldrich
Rotenone, ≥95%
Sigma-Aldrich
L-Glutathione reduced, ≥98.0%
Sigma-Aldrich
L-Glutathione reduced, Vetec, reagent grade, ≥98%
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Sod2
Sigma-Aldrich
HEPES, BioUltra, for molecular biology, ≥99.5% (T)
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
Carbonyl cyanide 4-(trifluoromethoxy)phenylhydrazone, ≥98% (TLC), powder
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Supelco
DL-Dithiothreitol solution, 1 M in H2O
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
Calcein-AM, Small Package (20 X 50 μg ), ≥90.0% (HPLC)
Sigma-Aldrich
Calcein-AM, suitable for fluorescence, BioReagent, ≥90% (HPLC)
SAFC
HEPES
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%
SAFC
HEPES