Merck
  • Biothiol-triggered, self-disassembled silica nanobeads for intracellular drug delivery.
Biothiol-triggered, self-disassembled silica nanobeads for intracellular drug delivery.
Acta biomaterialia (2015-05-20)
Xin-Chun Huang, Li-Bang Wu, Jen-Fang Hsu, Shinsuke Shigeto, Hsin-Yun Hsu
ABSTRACT
Silica-based nanomaterials have demonstrated great potential in biomedical applications due to their chemical inertness. However, the degradability and endosomal trapping issues remain as rate-limiting barriers during their innovation. In this study, we provide a simple yet novel sol-gel approach to construct the redox-responsive silica nanobeads (ReSiNs), which could be rapidly disassembled upon redox gradient for intracellular drug delivery. The disulfide-linked scaffold of the nanobead was synthesized by employing the dithiobis-(succinimidyl propionate) to bridge (3-aminopropyl)-trimethoxysilane. Such silica matrix could be efficiently disrupted in response to intracellular glutathione, resulting in drug release and collapse of entire nanocarrier. Moreover, the ReSiNs exhibited insignificant cytotoxicity before and after the degradation. These results indicated the potential of using ReSiNs as a novel silica-based, biothiol-degradable nanoplatform for future drug delivery.
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC
Sigma-Aldrich
5-Carboxy-tetramethylrhodamine N-succinimidyl ester, BioReagent, suitable for fluorescence
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
3,3′-Dithiodipropionic acid di(N-hydroxysuccinimide ester), powder
Sigma-Aldrich
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, for molecular biology
Sigma-Aldrich
Potassium hydroxide, anhydrous, ≥99.95% trace metals basis
Sigma-Aldrich
(3-Mercaptopropyl)trimethoxysilane, 95%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
(3-Aminopropyl)trimethoxysilane, 97%
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Supelco
Ethanol standards 10% (v/v), 10 % (v/v) in H2O, analytical standard