Merck
  • Effect of technical cashew nut shell liquid on rumen methane emission and lactation performance of dairy cows.
Effect of technical cashew nut shell liquid on rumen methane emission and lactation performance of dairy cows.
Journal of dairy science (2015-03-22)
A F Branco, F Giallongo, T Frederick, H Weeks, J Oh, A N Hristov
ABSTRACT
Technical-grade cashew nut shell liquid (TCNSL) is a by-product of the cashew nut industry in tropical countries, and is known to exhibit a wide range of biological activities, including inhibitory effect against gram-positive bacteria. This study was conducted to investigate the effects of TCNSL (73.3% cardanol, 16.4% cardol, and 3.0% methylcardol) on rumen methane emission, nutrient digestibility, dry matter intake, and milk yield and composition in dairy cows. Eight multiparous Holstein cows were used in a crossover design trial with two 21-d experimental periods. The diet was based on corn silage and alfalfa haylage and was formulated to meet or exceed the energy and metabolizable protein requirements of the cows. Treatments were control (no TCNSL supplementation) or 30 g/cow per day of TCNSL. Rumen carbon dioxide emission was not affected by TCNSL. Treatment had no effect on methane emission (542 vs. 511±35.3 g/cow per day, respectively) and methane emission intensity (15.0 vs. 13.9±0.58 g/kg of energy-corrected milk, respectively) and tended to decrease methane emission per kilogram of dry matter intake (20.2 vs. 18.6±1.04 g/kg, respectively). Dry matter intake (average 26.9±1.00 kg/d), milk yield (40.0±1.73 kg/d), and milk composition were not different between treatments. The TCNSL had no effect on N losses in urine and feces and total-tract apparent digestibility of nutrients, except digestibility of neutral detergent fiber tended to be increased compared with the control. Plasma urea and glucose concentrations were not affected by TCNSL. Concentration of milk C18:0 tended to be decreased (17%) by TCNSL compared with the control. In this study, TCNSL did not alter absolute methane emission in the rumen, but tended to decrease it by 8% per kilogram of dry matter intake. The TCNSL had no effect on milk yield and composition in dairy cows.
Sigma-Aldrich
Sodium sulfite, BioUltra, anhydrous, ≥98% (RT)
Sigma-Aldrich
Carbon dioxide, ≥99.8%
Sigma-Aldrich
Methane, electronic grade, ≥99.998%
Sigma-Aldrich
2-Thiobarbituric acid, ≥98%
Sigma-Aldrich
Sodium sulfite, BioXtra, ≥98%
Sigma-Aldrich
Carbon, mesoporous, nanopowder, less than 500 ppm Al, Ti, Fe, Ni, Cu, and Zn combined
Sigma-Aldrich
Carbon, mesoporous, nanopowder, graphitized, less than 250 ppm Al, Ti, Fe, Ni, Cu, and Zn combined
Sigma-Aldrich
Carbon, mesoporous, less than 100 ppm Al, Ti, Fe, Ni, Cu, and Zn combined
Sigma-Aldrich
Carbon, mesoporous
Sigma-Aldrich
Carbon, mesoporous, hydrophilic pore surface
Sigma-Aldrich
Carbon-12C dioxide, 99.9 atom % 12C
Sigma-Aldrich
Nitrogen, ≥99.998%
Sigma-Aldrich
Methane-12C, 13C-depleted, 99.9 atom % 12C
Sigma-Aldrich
Carbon nanofibers, pyrolitically stripped, platelets(conical), >98% carbon basis, D × L 100 nm × 20-200 μm
Sigma-Aldrich
Carbon-12C dioxide, 99.99 atom % 12C
Carbon - Vitreous, rod, 100mm, diameter 7.0mm, glassy carbon
Carbon - Vitreous, foil, 100x100mm, thickness 1.0mm, glassy carbon
Carbon - Vitreous, rod, 100mm, diameter 3.0mm, glassy carbon
Carbon - Vitreous, rod, 200mm, diameter 1.0mm, glassy carbon
Carbon - Vitreous, tube, 100mm, outside diameter 10mm, inside diameter 3mm, wall thickness 3.5mm, glassy carbon
Carbon - Vitreous, foil, 25x25mm, thickness 4.0mm, glassy carbon
Carbon - Vitreous, rod, 100mm, diameter 5.0mm, glassy carbon
Carbon - Vitreous, foam, 300x300mm, thickness 30mm, bulk density 0.05g/cm3, porosity 96.5%
Carbon - Vitreous, foam, 300x300mm, thickness 20mm, bulk density 0.05g/cm3, porosity 96.5%
Carbon - Vitreous, foil, 50x50mm, thickness 4.0mm, glassy carbon
Carbon - Vitreous, foil, 100x100mm, thickness 2.0mm, glassy carbon
Carbon - Vitreous, rod, 200mm, diameter 3.0mm, glassy carbon
Carbon - Vitreous, rod, 200mm, diameter 5.0mm, glassy carbon
Carbon - Vitreous, rod, 5mm, diameter 3.0mm, glassy carbon
Carbon - Vitreous, foil, 25x25mm, thickness 0.5mm, glassy carbon