Merck
  • Selective oxidative stress and cholesterol metabolism in lipid-metabolizing cell classes: Distinct regulatory roles for pro-oxidants and antioxidants.
Selective oxidative stress and cholesterol metabolism in lipid-metabolizing cell classes: Distinct regulatory roles for pro-oxidants and antioxidants.
BioFactors (Oxford, England) (2015-08-01)
Mira Rosenblat, Nina Volkova, Michael Aviram
ABSTRACT
Atherogenesis is associated with macrophage cholesterol and oxidized lipids accumulation and foam cell formation. However, two other major lipid-metabolizing cell classes, namely intestinal and liver cells, are also associated with atherogenesis. This study demonstrates that manipulations of cellular oxidative stress (by fatty acids, glucose, low-density lipoprotein, angiotensin II, polyphenolic antioxidants, or the glutathione/paraoxonase 1 systems) have some similar, but also some different effects on cholesterol metabolism in macrophages (J774A.1) versus intestinal cells (HT-29) versus liver cells (HuH7). Cellular oxidative stress was ≈3.5-folds higher in both intestinal and liver cells versus macrophages. In intestinal cells or liver cells versus macrophages, the cholesterol biosynthesis rate was increased by 9- or 15-fold, respectively. In both macrophages and intestinal cells C-18:1 and C-18:2 but not C-18:0, fatty acids significantly increased oxidative stress, whereas in liver cells oxidative stress was significantly decreased by all three fatty acids. In liver cells, trans C-18:1 versus cis C-18:1, unlike intestinal cells or macrophages, significantly increased cellular oxidative stress and cellular cholesterol biosynthesis rate. Pomegranate juice (PJ), red wine, or their phenolics gallic acids or quercetin significantly reduced cellular oxidation mostly in macrophages. Recombinant PON1 significantly decreased macrophage (but not the other cells) oxidative stress by ≈30%. We conclude that cellular atherogenesis research should look at atherogenicity, not only in macrophages but also in intestinal and liver cells, to advance our understanding of the complicated mechanisms behind atherogenesis. © 2015 BioFactors, 41(4):273-288, 2015.
Sigma-Aldrich
Sodium hydroxide-16O solution, 20 wt. % in H216O, 99.9 atom % 16O
Sigma-Aldrich
Nitrogen, ≥99.998%
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Oleic acid, meets analytical specification of Ph, Eur., 65.0-88.0% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
2-Propanol, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
2-Propanol, BioReagent, for molecular biology, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Oleic acid, BioReagent, suitable for cell culture
Sigma-Aldrich
L-Glutathione reduced, ≥98.0%
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
SAFC
Cholesterol, Plant-Derived, SyntheChol®
Sigma-Aldrich
2-Propanol, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Oleic acid, natural, FCC
Sigma-Aldrich
Oleic acid, technical grade, 90%
Sigma-Aldrich
L-Glutathione reduced, Vetec, reagent grade, ≥98%
Sigma-Aldrich
Dimethyl sulfoxide, Vetec, reagent grade, 99%
Sigma-Aldrich
Stearic acid, Grade I, ≥98.5% (capillary GC)
Sigma-Aldrich
Gallic acid, 97.5-102.5% (titration)
Sigma-Aldrich
Stearic acid, ≥95%, FCC, FG
Sigma-Aldrich
Hexane, HPLC Plus, for HPLC, GC, and residue analysis, ≥95%
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Oleic acid, ≥99% (GC)
Sigma-Aldrich
Stearic acid, reagent grade, 95%
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
L-Glutathione reduced, BioXtra, ≥98.0%
Sigma-Aldrich
L-Glutathione reduced, suitable for cell culture, BioReagent, ≥98.0%, powder