Merck
  • ATF4 Gene Network Mediates Cellular Response to the Anticancer PAD Inhibitor YW3-56 in Triple-Negative Breast Cancer Cells.
ATF4 Gene Network Mediates Cellular Response to the Anticancer PAD Inhibitor YW3-56 in Triple-Negative Breast Cancer Cells.
Molecular cancer therapeutics (2015-01-24)
Shu Wang, Xiangyun Amy Chen, Jing Hu, Jian-Kang Jiang, Yunfei Li, Ka Yim Chan-Salis, Ying Gu, Gong Chen, Craig Thomas, B Franklin Pugh, Yanming Wang
ABSTRACT
We previously reported that a pan-PAD inhibitor, YW3-56, activates p53 target genes to inhibit cancer growth. However, the p53-independent anticancer activity and molecular mechanisms of YW3-56 remain largely elusive. Here, gene expression analyses found that ATF4 target genes involved in endoplasmic reticulum (ER) stress response were activated by YW3-56. Depletion of ATF4 greatly attenuated YW3-56-mediated activation of the mTORC1 regulatory genes SESN2 and DDIT4. Using the ChIP-exo method, high-resolution genomic binding sites of ATF4 and CEBPB responsive to YW3-56 treatment were generated. In human breast cancer cells, YW3-56-mediated cell death features mitochondria depletion and autophagy perturbation. Moreover, YW3-56 treatment effectively inhibits the growth of triple-negative breast cancer xenograft tumors in nude mice. Taken together, we unveiled the anticancer mechanisms and therapeutic potentials of the pan-PAD inhibitor YW3-56.
Sigma-Aldrich
MISSION® esiRNA, targeting human ATF4
Sigma-Aldrich
Formaldehyde-12C solution, 20% in H2O, 99.9 atom % 12C
Sigma-Aldrich
MISSION® esiRNA, targeting human EIF2AK3, AC104134.2
Sigma-Aldrich
Formaldehyde solution, for molecular biology, 36.5-38% in H2O
Sigma-Aldrich
Calcium Ionophore A23187, ≥98% (TLC), powder
Sigma-Aldrich
Formaldehyde solution, meets analytical specification of USP, ≥34.5 wt. %
Sigma-Aldrich
Formaldehyde solution, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Sigma-Aldrich
Formaldehyde solution, for molecular biology, BioReagent, ≥36.0% in H2O (T)
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Ddit4
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Eif2ak3
Sigma-Aldrich
MISSION® esiRNA, targeting human SESN2
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Sesn2
Sigma-Aldrich
Hematoxylin, certified by the Biological Stain Commission
Sigma-Aldrich
Hematoxylin
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Atf4
Sigma-Aldrich
MISSION® esiRNA, targeting human DDIT4
Sigma-Aldrich
Anti-β-Actin antibody, Mouse monoclonal, clone AC-15, purified from hybridoma cell culture