Merck
  • Structural insights into the benzophenanthridines binding to human glycine transporter GlyT1.
Structural insights into the benzophenanthridines binding to human glycine transporter GlyT1.
European journal of pharmacology (2015-08-15)
Frantisek Jursky, Martina Baliova, Anna Juhasova
ABSTRACT
We previously identified cysteine 475 as a key residue for the inhibitory action of sanguinarine on the human glycine transporter GlyT1c. To define potential benzophenanthridine binding pocket more closely, we created a structural homology model of GlyT1 and also mutated several amino acids in the vicinity of cysteine 475. Even though this area contains the molecular determinants of the glycine and sodium permeation pathways, and several mutations resulted in an inactive transporter, we found that the mutation of a polar aromatic tyrosine 370 to purely aromatic phenylalanine, but not to an aliphatic leucine, significantly increased the sensitivity of GlyT1 to both sanguinarine and chelerythrine. The reduction of sanguinarine to dihydrosanguinarine completely eliminated the alkaloid's inhibitory potency. Both these results suggest that aromaticity is important in the interaction of benzophenanthridines with GlyT1. Even though tyrosine 370 is part of the conformationaly highly flexible glycine binding site, and is accesible during the transport process from both intra and extracellular sites, the cytoplasmic location of the second alkaloid sensitive residue, cysteine 475, suggests that the benzophenanthridines might attack the area of the GlyT1 intracellular gates.
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Trizma® base, ≥99.0% (T)
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
2-Mercaptoethanol, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Trizma® base, BioUltra, for molecular biology, ≥99.8% (T)
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Tris(hydroxymethyl)aminomethane, ACS reagent, ≥99.8%
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Lithium-7Li chloride, 99 atom % 7Li, 99% (CP)
Sigma-Aldrich
Lithium chloride, AnhydroBeads, −10 mesh, ≥99.9% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Lithium chloride, AnhydroBeads, −10 mesh, 99.998% trace metals basis
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Lithium chloride, powder, ≥99.98% trace metals basis
Sigma-Aldrich
2-Mercaptoethanol, ≥99.0%
Sigma-Aldrich
p-Coumaric acid, ≥98.0% (HPLC)
Sigma-Aldrich
Lithium chloride, for molecular biology, ≥99%
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Lithium chloride solution, 8 M, for molecular biology, ≥99%
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Trizma® base, ≥99.9% (titration), crystalline
Sigma-Aldrich
Trizma® base, BioXtra, pH 10.5-12.0 (1 M in H2O), ≥99.9% (titration)
Sigma-Aldrich
2-Mercaptoethanol, for molecular biology, suitable for electrophoresis, suitable for cell culture, BioReagent, 99% (GC/titration)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Lithium chloride, BioXtra, ≥99.0% (titration)